Skip to Content
Merck

Skip To

MABE324

Anti-MSH3 Antibody, clone 1F6

clone 1F6, 1 mg/mL, from mouse

Synonym(s):

DNA mismatch repair protein Msh3, Protein repair-1, REP-1, Protein repair-3, REP-3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 401,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
1F6, monoclonal
Application:
ICC, WB
Citations:
-

€ 401,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F6, monoclonal

species reactivity

mouse

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Msh3(17686)

General description

MSH3, also known as DNA mismatch repair protein Msh3, Protein repair-1, REP-1, or Protein repair-3, REP-3, and encoded by the gene MSH3/REP-3, is an important component of the post-replicative DNA mismatch repair system. MSH3 heterodimerizes with MSH2 to form a complex named MutS beta which binds DNA mismatches and initiates DNA repair. MSH3/MSH2 complex (MutS beta) also complexes with another protein complex called MutL alpha which is also necessary for further steps in DNA repair. MSH3 is also frequently mutated in many cancers resulting in a mismatch repair deficiency and this can contribute to sensitivity to cytotoxic drugs. Interestingly too, because MSH3 is involved in DNA repair, it appears to it can mediate sensitivity to various repeat-associated diseases such as Huntington’s disease.
~123 kDa observed

Immunogen

Recombinant protein corresponding to mouse MSH3.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
This Anti-MSH3 Antibody, clone 1F6 is validated for use in Western Blotting and Immunocytochemistry for the detection of MSH3.
Western Blotting Analysis: 0.5 µg/mL of this antibody detected MSH3 in 10 µg of mouse testis tissue lysate.

Immunocytochemistry Analysis: A representative lot detected MSH3 in C2C12 cells (Prof. Glenn Morris, Wolfson Centre for Inherited Neuromuscular Disease.; Holt, I., et al. (2011). J Cell Biochem. 112(6):1612-1621.).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in C2C12 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected MSH3 in 10 µg of C2C12 cell lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4501185SAB4501333SAB5600264
Gene Information

mouse ... Msh3(17686)

Gene Information

human ... MLH3(27030)

Gene Information

human ... MSH6(2956)

Gene Information

human ... MSH2(4436)

clone

1F6, monoclonal

clone

polyclonal

clone

polyclonal

clone

RM375, monoclonal, recombinant monoclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

species reactivity

mouse

species reactivity

human

species reactivity

human, mouse

species reactivity

human

biological source

mouse

biological source

rabbit

biological source

rabbit

biological source

-


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters