Skip to Content
Merck

Skip To

MABE308

Anti-ICBP90/UHRF1 Antibody, clone 1RC1C-10

ascites fluid, clone 1RC-1C10, from mouse

Synonym(s):

E3 ubiquitin-protein ligase, inverted CCAAT box-binding protein, Nuclear protein 95, Nuclear zinc finger protein Np95, HuNp95, RING finger protein 106, Transcription factor ICBP90, Ubiquitin-like PHD and RING finger domain-containing protein 1, Ubiquitin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Check Cart for Availability
€ 467,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
1RC-1C10, monoclonal
Application:
ChIP, ICC, IHC, WB
Citations:
6

€ 467,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1RC-1C10, monoclonal

species reactivity

human

technique(s)

ChIP: suitable, immunocytochemistry: suitable, immunohistochemistry: suitable, western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... UHRF1(29128)

General description

Ubiquitin-like with PHD and ring finger domains 1 (UHRF1), also known as ICBP90 or Np95, is a multifunctional nuclear protein that may be an important mediator of heterochromatin formation which is indicative of gene repression. UHRF1 contains a set and RING-finger-associated domain that detects hemi-methylated cytosine residues leading to the recruitment of DNMT1 to these residues during the S phase. UHRF1 thereby facilitates the transfer of methylation status to newly synthesized DNA molecules. UHRF1 may also interact with histones and HDAC1 via its PHD domain to promote modifications of histone residues which contribute to heterochromatin formation. UHRF1 may also directly modify histone H3 by ubiquitination. In addition, UHRF1 may contribute to the DNA-repair process though associations with PCNA. Several studies have suggested that overexpression of UHRF1 may contribute to the development of many cancers.
~90 kDa observed

Immunogen

Recombinant protein corresponding to human ICBP90.

Application

Anti-ICBP90/UHRF1 Antibody, clone 1RC1C-10 is a Mouse Monoclonal Antibody for detection of ICBP90/UHRF1 also known as E3 ubiquitin-protein ligase, inverted CCAAT box-binding protein & has been validated in WB, ChIP, ICC & IHC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
Western Blot Analysis: A representative lot from an independent laboratory detected ICBP90/UHRF1 in HeLa and MOLT-4 cell lysates (Hopfner, R., et al. (2001). 266(1-2)15-23.).

Immunocytochemistry Analyisis: A representative lot from an independent laboratory detected ICBP90/UHRF1 in ICC (Hopfner, R., et al. (2001). Gene. 266(1-2)15-23.).

Immunohistochemistry Analysis: A representative lot from an independent laboratory detected ICBP90/UHRF1 in human appendix and human breast carcinoma tissues (Hopfner, R., et al. (2000). Cancer Res. 60(1):121-128.).

Chromatin Immunoprecipitation Analyis: A representative lot from an independent laboratory immunoprecipitated ICB90/UHRF1 in ChIP (Felle, M., et al. (2011). Nucleic Acids Res. 39(19). 8355-8365.)

Physical form

Mouse monoclonal IgG1 ascites with 0.05% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa cell lysate
Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: A 1:5,000 dilution of this antibody detected ICBP90/UHRF1 in 10 µg of HeLa cell lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABE945WH0029128M1SAB1405050
biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

clone

1RC-1C10, monoclonal

clone

7C8, monoclonal

clone

3A11, monoclonal

clone

3B12, monoclonal

Gene Information

human ... UHRF1(29128)

Gene Information

human ... UHRF1(29128)

Gene Information

human ... UHRF1(29128)

Gene Information

human ... UHRF1(29128)

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

antibody form

ascites fluid

antibody form

purified antibody

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters