Skip to Content
Merck
All Photos(1)

Key Documents

MABE306

Sigma-Aldrich

Anti-Dnmt1 Antibody, clone DNM-2C1

clone DNM-2C1, from rat

Synonym(s):

DNA (cytosine-5)-methyltransferase 1, Dnmt1, CXXC-type zinc finger protein 9, DNA methyltransferase HsaI, DNA Mtase Hsal, M. Hsal, MCMT

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€487.00

€487.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€487.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€487.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

DNM-2C1, monoclonal

species reactivity

human

technique(s)

ChIP: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DNMT1(1786)

General description

DNA (cytosine-5)-methyltransferase 1 (Dnmt1) is one of three mammalian enzymes that catalyze the addition of a methyl group to the 5’ carbon of cytosine residues in CpG dinucleotides. These methylation marks are heritable modifications that are transferred to new DNA molecules following replication, by the activity of Dnmt1, and possibly Dnmt3a and Dnmt3b. Dnmt1 recognizes hemimethylated CpG regions with the aid of the UHRF1 protein, and it mediates the methylation of cytosines at those sites. DNA methylation has been linked to chromosome stability and gene expression in somatic cells; and to parental imprinting and X-chromosome inactivation in germ cells. Several studies have indicated that DNA methylation patterns are altered in various cancers.

Immunogen

His-tagged recombinant protein corresponding to human Dnmt1.

Application

Anti-Dnmt1 Antibody, clone DNM-2C1 is a Rat Monoclonal Antibody for detection of Dnmt1 also known as DNA (cytosine-5)-methyltransferase 1 & has been validated in IP, ChIP & WB.
Immunoprecipitation Analysis: A representative lot was used by an independent laboratory to detect Dnmt1 in IP (Felle, M., et al. (2011). Nucleic Acids Res. 39(19):8355-8365.).

Chromatin Immunoprecipitation Analysis: A representative lot was used by an independent laboratory to immunoprecipitate Dnmt1 in ChIP (Felle, M., et al. (2011). Nucleic Acids Res. 39(19):8355-8365.).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Western Blot in HeLa nuclear extract.

Western Blot Analysis: 0.02 µg/mL of this antibody detected Dnmt1 in 10 µg of HeLa nuclear extract.

Target description

~183 kDa observed. Three isoforms are known to exist at ~183 kDa, ~185 kDa, and ~145 kDa An uncharacterized band may be observed at ~90 kDa in some nuclear extracts.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa nuclear extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher Grunseich et al.
Molecular cell, 69(3), 426-437 (2018-02-06)
R-loops are three-stranded nucleic acid structures found abundantly and yet often viewed as by-products of transcription. Studying cells from patients with a motor neuron disease (amyotrophic lateral sclerosis 4 [ALS4]) caused by a mutation in senataxin, we uncovered how R-loops

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service