Skip to Content
Merck
All Photos(4)

Key Documents

MABE263

Sigma-Aldrich

Anti- Histone H3c1 Antibody

mouse monoclonal, 9B1-2G6

Synonym(s):

Histone H3.1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€408.00

€408.00


Estimated to ship on30 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€408.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€408.00


Estimated to ship on30 April 2025


Request a Bulk Order

Product Name

Anti-unmethylated-Histone H3 (Lys9) Antibody, clone 9B1-2G6, clone 9B1-2G6, from mouse

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

9B1-2G6, monoclonal

species reactivity

human

technique(s)

dot blot: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... H3C1(8350)

General description

Histones are nuclear proteins that form octameric structures which bind DNA to form units of chromatin called nucleosomes. The family of histones—H2A, H2B, H3, and H4—are key players in gene regulation. They undergo a number of post-translational modifications (PTM) in response to various stimuli, including phosphorylation on serine and threonine residues and methylation on lysine residues. PTMs produce configural changes in histone proteins that may induce nucleosome remodeling and expose or hide DNA sequences from transcriptional complexes. Methylation occurs on histone H3 at lysine 4, 9, 27, 36, and 79 residues. Previous studies have reported that methylation of H3 lysine 9 provides a binding site for heterochromatin adaptor proteins (HP1) which are involved in the formation of heterochromatin and gene repression. Conversely,deacetylation of histone H3 lysine 9 is associated with gene-activating events. Often, it is the pattern of different modifications on several histone residues that alter chromatin, rather than modification of single residues.

Specificity

This antibody recognizes Histone H3 unmethylated at Lys9. This antibody does not cross react with methyl-Histone (Lys29). This antibody detects unmethylated Histone H3 (Lys9) when Ser10 is phosphorylated. It also detects mono-/di-methyl-Histone H3 (Lys4), and Acetyl-Histone H3 (Lys14). (Prof. E. Ogris, Department of Medical Biochemistry, Medical University of Vienna).

Immunogen

Epitope: unmethylated Lys9
KLH-conjugated linear peptide corresponding to human unmethylated Histone H3 (Lys9).

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-unmethylated-Histone H3 (Lys9) Antibody, clone 9B1-2G6 (Mouse Monoclonal Antibody) demonstrated to detect unmethylated-Histone H3 (Lys9) also known as Histone H3.1.
Western Blot Analysis: 0.5 µg/mL from a representative lot detected unmethylated Histone H3 (Lys9) in 10 µg of HeLa acid extract.

Dot Blot Analysis: Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) and Absurance Histone H2A, H2B, H4 Antibody Specificity Array (Cat. No. 16-665), which contain histone peptides with various modifications were probed with Cat. No MABE263-S, unmethylated-Histone H3 (Lys9), clone 9B1-2G6 at 0.5 µg/mL (1:2,000) dilution. Proteins were visualized using a Donkey anti-rabbit IgG conjugated to HRP and a chemiluminescence detection system.

Dot Blot (Specificity) Analysis: A representative lot detected non-methylated Histone H3 (Lys9) in Dot Blot. This antibody detects non-methylated Histone H3 (Lys9) when Ser10 is phosphorylated. It also detects mono-/di-methyl-Histone H3(Lys4), and Acetyl-Histone H3 (Lys14). (Prof. E. Ogris, Department of Medical Biochemistry, Medical University of Vienna).

Quality

Evaluated by Western Blot in recombinant Histone H3.

Western Blot Analysis: 0.5 µg/mL of this antibody detected unmethylated Histone H3 (Lys9) in 10 µg of recombinant Histone H3.

Target description

~15 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ supernatant in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Recombinant Histone H3

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yichen Ge et al.
Toxicology and applied pharmacology, 355, 164-173 (2018-07-04)
Arsenic is a known potent risk factor for bladder cancer. Increasing evidence suggests that epigenetic alterations, e.g., DNA methylation and histones posttranslational modifications (PTMs), contribute to arsenic carcinogenesis. Our previous studies have demonstrated that exposure of human urothelial cells (UROtsa

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service