Skip to Content
Merck

Skip To

MABE253

Anti-Ago2 Antibody, clone 11A9

clone 11A9, from rat

Synonym(s):

Protein argonaute-2, Argonaute2, hAgo2, Eukaryotic translation initiation factor 2C 2, eIF-2C 2, eIF2C 2, PAZ Piwi domain protein, PPD, Protein slicer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
200 μL
Check Cart for Availability
€ 428,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
11A9, monoclonal
Application:
ICC, IP, WB
Citations:
31

€ 428,00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1205

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

11A9, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AGO2(27161)

General description

Ago2 (argonaute-2), also known as eukaryotic translation initiation factor 2C (EIF2C2), is an essential component for siRNA-directed RNA interference (RNAi) response. Ago2 is an endonuclease required for the unwinding of siRNA duplex and assembly of siRNA into RNA-induced silencing complexes (RISC). Ago2 interacts with DICER1 through its Piwi domain. This Piwi domain is thought to provide RNA cleavage activity via a mechanism similar to RNase H. Ago2 activity is necessary for embryonic development as well as RNA-mediated gene silencing (RNAi).
~85 kDa observed. Two isoforms at ~97 kDa and ~94 kDa are known to exist.

Immunogen

Linear peptide corresponding to human Ago2.

Application

Immunoprecipitation Analysis: A representative lot from an independent laboratory detected Ago2 in HEK239 cell lysate (Rudel, S., et al. (2008) RNA. 14(6):1244-1253.).

Immunocytochemistry Analysis: A represeentative lot from an independent laboratory detected Ago2 in HEK293, HeLa, and RPE-1 cells (Weinmann, L., et al. (2009). Cell. 136(3):496-507.).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins
Use Anti-Ago2 Antibody, clone 11A9 (Rat Monoclonal Antibody) validated in WB, IP, ICC to detect Ago2 also known as Protein argonaute-2, Argonaute2, hAgo2, eIF-2C 2.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate
Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: A 1:500 dilution of this antibody detected Ago2 in 10 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4200724MABE144SAB1306339
antibody form

purified antibody

antibody form

purified from hybridoma cell culture

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

clone

11A9, monoclonal

clone

AGO2-10, monoclonal

clone

4B1-F6, monoclonal

clone

polyclonal

Gene Information

human ... AGO2(27161)

Gene Information

human ... AGO2(27161)

Gene Information

human ... AGO3(192669)

Gene Information

human ... AGO1(26523)

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

biological source

rat

biological source

mouse

biological source

mouse

biological source

rabbit


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters