Skip to Content
Merck
All Photos(2)

Key Documents

MABE191

Sigma-Aldrich

Anti-WHSC1/NSD2 Antibody, clone 29D1

clone 29D1, from mouse

Synonym(s):

Probable histone-lysine N-methyltransferase NSD2, Multiple myeloma SET domain-containing protein, Nuclear SET domain-containing protein 2, Protein trithorax-5, Wolf-Hirschhorn syndrome candidate 1 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€432.00

€432.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€432.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€432.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

29D1, monoclonal

species reactivity

human

technique(s)

ChIP: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WHSC1(7468)

General description

Wolf-Hirschhorn syndrome candidate 1 protein (WHSC1), Nuclear SET domain containing protein 2 (NSD2) is a histone lysine methyltransferase that is deregulated in a subgroup of multiple myelomas. NSD proteins have been described to serve as oncogenes; however, little is known about their targets in transcriptional repression or activation. NSD2 is overexpressed in multiple myelomas via translocation between the NSD2 locus on chromosome 4 and the immunoglobulin locus on chromosome 14. NSD2 is under the regulatory control of this immunoglobulin locus. Further research is required to elucidate its roles and mechanisms in cancer.

Immunogen

GST-tagged recombinant protein corresponding to human WHSC1/NSD2.

Application

Detect WHSC1/NSD2 using this Anti-WHSC1/NSD2 Antibody, clone 29D1 validated for use in Western Blotting, Chromatin Immunoprecipitation (ChIP), Immunoprecipitation.
Western Blot Analysis: 0.5-1 µg/mL from a representative lot detected WHSC1/NSD2 in 10 µg of HeLa nuclear extract and human placenta tissue lysate.

Chromatin Immunoprecipitation Analysis: A representative lot was used by an independent laboratory to detect WHSC1/NSD2 in transfected Gal4-reporter/293T cells. (Marango, J., et al. (2008). Blood. 111:3145-3154.)

Immunoprecipitation Analysis: A representative lot was used by an independent laboratory to detect WHSC1/NSD2 in KMS11 cells. (Marango, J., et al. (2008). Blood. 111:3145-3154.)

Quality

Evaluated by Western Blot in HCT116 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected WHSC1/NSD2 in 10 µg of HCT116 cell lysate.

Target description

~152 kDa observed. There are seven isoforms ranging between ~30 and ~152 kDa Different lysates may indicate different isoforms in Western Blots.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon Papillon-Cavanagh et al.
Nature genetics, 49(2), 180-185 (2017-01-10)
Human papillomavirus (HPV)-negative head and neck squamous cell carcinomas (HNSCCs) are deadly and common cancers. Recent genomic studies implicate multiple genetic pathways, including cell signaling, cell cycle and immune evasion, in their development. Here we analyze public data sets and
Jia-Ray Yu et al.
Science advances, 7(29) (2021-07-16)
Histone H3K27M is a driving mutation in diffuse intrinsic pontine glioma (DIPG), a deadly pediatric brain tumor. H3K27M reshapes the epigenome through a global inhibition of PRC2 catalytic activity and displacement of H3K27me2/3, promoting oncogenesis of DIPG. As a consequence
Salina Yuan et al.
Cancer discovery, 10(6), 854-871 (2020-03-20)
Epithelial plasticity, reversible modulation of a cell's epithelial and mesenchymal features, is associated with tumor metastasis and chemoresistance, leading causes of cancer mortality. Although different master transcription factors and epigenetic modifiers have been implicated in this process in various contexts
Ashot S Harutyunyan et al.
Cell reports, 33(7), 108390-108390 (2020-11-19)
The discovery of H3K27M mutations in pediatric gliomas marked a new chapter in cancer epigenomics. Numerous studies have investigated the effect of this mutation on H3K27 trimethylation, but only recently have we started to realize its additional effects on the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service