Select a Size
About This Item
biological source
rat
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
4F5, monoclonal
mol wt
calculated mol wt 155.14 kDa
observed mol wt ~155 kDa
purified by
using protein G
species reactivity
human
packaging
antibody small pack of 100 μL
technique(s)
ELISA: suitable
immunofluorescence: suitable
western blot: suitable
isotype
IgG2aκ
epitope sequence
C-terminal half
Protein ID accession no.
UniProt accession no.
shipped in
2-8°C
target post-translational modification
unmodified
Gene Information
human ... AKNA(80709)
Related Categories
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in HeLa cell lysate.
Western Blotting Analysis: A 1:500 dilution of this antibody detected AKNA in HeLa cell lysate.
Tested Applications
Immunofluorescence Analysis: A representative lot detected AKNA in Immunofluorescence applications (Camargo Ortega, G., et al. (2019). Nature. 567(7746):113-117).
ELISA Analysis: A representative lot detected AKNA in ELISA applications (Camargo Ortega, G., et al. (2019). Nature. 567(7746):113-117).
Western Blotting Analysis: A representative lot detected AKNA in Western Blotting applications (Camargo Ortega, G., et al. (2019). Nature. 567(7746):113-117).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service