Skip to Content
Merck
All Photos(4)

Key Documents

MABE1796M

Sigma-Aldrich

Anti-YL-1 Antibody, clone EX-1Y32

clone EX-1Y32, from mouse

Synonym(s):

Vacuolar protein sorting-associated protein 72 homolog, Protein YL-1, Transcription factor-like 1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€116.00
100 μG
€282.00

€116.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€116.00
100 μG
€282.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€116.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

EX-1Y32, monoclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

General description

Vacuolar protein sorting-associated protein 72 homolog (UniProt: Q15906; also known as Protein YL-1, Transcription factor-like 1) is encoded by the VPS72 (also known as TCFL1, YL1) gene (Gene ID: 6944) in human. YL-1 is a nuclear protein that may serve as a DNA-binding transcriptional regulator and may be involved in chromatin modification and remodeling. YL-1 is plays multiple roles in chromatin modification and remodeling in cells and it is shown to be a subunit of TRRAP/TIP60 HAT complex and of chromatin remodeling SCCAP-containing complex. It has a DNA binding region that includes amino acids 156 to 206. In addition, it has an aspartic/glutamic acid rich acidic region (aa 22-77) and a proline rich region (aa 333-354). Two isoforms of YL-1 have been described that are produced by alternative splicing. (Ref.: Cai, Y., et al. (2006). J. Biol. Chem. 280 (14); 13665-70).

Specificity

Clone EX-1Y32 detects Vacuolar protein sorting-associated protein 72 homolog (YL-1) in human and murine cells.

Immunogen

GST-tagged full-length recombinant human YL-1 protein.

Application

Anti-YL-1, clone EX-1Y32, Cat. No. MABE1796, is a highly specific mouse monoclonal antibody that targets Vacuolar protein sorting-associated protein 72 homolog and has been tested for use in Immunocytochemistry, Immunoprecipitation, and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A representative lot detected YL-1 in Western Blotting applications (Obri, A., et. al. (2014). Nature. 505(7485):648-53).

Immunocytochemistry Analysis: A 1:800 dilution from a representative lot detected YL-1 in HeLa cells.

Western Blotting Analysis: 2 µg/mL from a representative lot detected YL-1 in 10 µg of HeLa cell lysate.

Immunoprecipitation Analysis: 1-10 µg from a representative lot detected YL-1 in Mouse embryonic fibroblast (MEF) total cell extracts.

Quality

Evaluated by Western Blotting in Mouse embryonic fibroblast (MEF) cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected YL-1 in Mouse embryonic fibroblast (MEF) cell lysate.

Target description

~48 kDa observed, 40.59 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein A + Protein G
Purified mouse monoclonal antibody IgG1 in PBS without azide and 40% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service