Skip to Content
Merck
All Photos(1)

Key Documents

MABE1795

Sigma-Aldrich

Anti-PP4C Antibody, clone 7B5

clone 7B5, from mouse

Synonym(s):

Serine/threonine-protein phosphatase 4 catalytic subunit, EC: 3.1.3.16, Pp4, PPP4C, Protein phosphatase X, PP-X

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€154.00
100 μL
€403.00

€154.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€154.00
100 μL
€403.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€154.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7B5, monoclonal

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), rat (based on 100% sequence homology), rabbit (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... PPP4C(5531)

General description

Serine/threonine-protein phosphatase 4 catalytic subunit (UniProt: P60510; also known as EC: 3.1.3.16, PP4C, Pp4, Protein phosphatase X, PP-X) is encoded by the PPP4C (also known as PPP4, PPX) gene (Gene ID: 5531) in human. Protein phosphatase 4 (PP4), a ubiquitous serine/threonine phosphatase, is involved in the regulation of many cellular processes. It functions independently of protein phosphatase 2A (PP2A). Mammalian PP4 contains two regulatory subunits (R1 and R2) that interact with the catalytic subunit (PP4C)) and control its activity. Methylation at the C-terminal Leucine 307 is critical for interactions with regulatory subunits. PP4C-R2 complexes are reported to be involved in organelle assembly, maturation of the centrosome, apoptosis, DNA repair, and spliceosomal assembly. It is also reported to play a role in cell migration and in activation of c-Jun N-terminal kinase and NF-kappa B. PP4C has multiple metal ion binding sites and can bind two manganese ions per subunit. (Ref.: Cohen, PTW., et al. (2005). FEBS Lett. 579(15); 3278-3286).

Specificity

Clone 7B5 specifically detects human Serine/threonine-protein phosphatase 4 catalytic subunit (PP4C). It targets an epitope within 17 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide coreesponding to 17 amino acids from the C-terminal region of human Serine/threonine-protein phosphatase 4 catalytic subunit (PP4C).

Application

Anti-PP4C, clone 7B5, Cat. No. MABE1795, is a highly specific mouse monoclonal antibody that targets Serine/threonine-protein phosphatase 4 catalytic subunit and has been tested for use in Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A 1:500 dilution from a representative lot detected PP4C in lysate from NIH3T3 mouse fibroblasts ectopically expressing N-terminally tagged human PP4 catalytic subunit (PPP4C).(Courtesy of Dr. Stefan Schuechner and Dr. Egon Ogris, Medical University of Vienna, Austria).

Quality

Isotype testing: Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgG1.

Target description

35.08 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service