Skip to Content
Merck
All Photos(1)

Key Documents

MABE1782

Sigma-Aldrich

Anti-phospho-E2F1 (Ser375) Antibody, clone 71-10

clone 71-10, from rabbit

Synonym(s):

Transcription factor E2F1, E2F-1, PBR3, Retinoblastoma-associated protein 1, RBAP-1, Retinoblastoma-binding protein 3, RBBP-3, pRB-binding protein E2F-1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€154.00
100 μL
€403.00

€154.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
€154.00
100 μL
€403.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€154.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

71-10, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

phosphorylation (pSer375/pSer375)

Gene Information

human ... E2F1(1869)

General description

Transcription factor E2F1 (UniProt: Q01094; also known as E2F-1, PBR3, Retinoblastoma-associated protein 1, RBAP-1, Retinoblastoma-binding protein 3, RBBP-3, pRB-binding protein E2F-1) is encoded by the E2F1 (also known as RBBP3) gene (Gene ID: 1869) in human. E2F-1 is a transcription activator that binds DNA cooperatively with DP proteins through the E2 recognition site, 5′-TTTC[CG]CGC-3′ found in the promoter region of a number of genes whose products are involved in cell cycle regulation or in DNA replication. It is a component of the DRTF1/E2F transcription factor complex and forms heterodimers with DP family members. The DRTF1/E2F complex functions in the control of cell-cycle progression from G1 to S phase. The E2F1 complex is shown to bind specifically hypophosphorylated retinoblastoma protein RB1. During the cell cycle, RB1 becomes phosphorylated in mid-to-late G1 phase and detaches from the DRTF1/E2F complex that renders E2F transcriptionally active. E2F-1 can be phosphorylated by CDK2 and cyclin A-CDK2 in the S-phase. It is also phosphorylated at serine 364 by CHEK2, which stabilizes it upon DNA damage and regulates its effect on transcription and apoptosis. Acetylation of E2F-1 by P/CAF/KAT2B stimulates its DNA-binding.

Specificity

Clone 71-10 is a rabbit monoclonal antibody that specifically detects human E2F1 phosphorylated on Serine 375.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids surrounding phosphoserine 375 from the C-terminal region of human transcription factor E2F1.

Application

Anti-phospho-E2F1 (Ser375), clone 71-10, Cat. No. MABE1782, is a highly specific rabbit monoclonal antibody that targets Transcription factor E2F1 phosphorylated on Serine 375 and has been tested in ELISA and Western Blotting.
ELISA Analysis: A representative lot detected phospho-E2F1 (Ser375) in ELISA applications (Dale, T., et. al. (2015). Nat Chem Biol. 11(12):973-980).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HEK293 cells transfected with pcDNA3.1 empty vector, HEK 293 cells transfected with E2F1 wild type-GST, HEK 293 cells transfected with E2F1 wild type-GST with alanine substituted for serine at 375.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected phospho-E2F1 (Ser375) in HEK293 cells transfected with E2F1 wild type-GST,, but not in HEK 293 cells transfected with E2F1 wild type-GST with alanine substituted for serine at 375.

Target description

~80 kDa observed ( GST fusion protein); 46.92 kDa calculated (without GST). Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rabbit monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service