Skip to Content
Merck
All Photos(2)

Key Documents

MABD94

Sigma-Aldrich

Anti-SOX-15 Antibody, clone 1A2.1

clone 1A2.1, from mouse

Synonym(s):

SOX-12, SOX-20, SOX-26, SOX-27

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€316.00

€316.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€316.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€316.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A2.1, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SOX12(6666)

General description

Members of the SOX family of transcription factors are widely conserved and play important roles in early development and are often used as markers to assess the differentiation of specific cell lineages. SOX-15 (also known as SOX-20) is a nuclear, 25 kDa transcriptional repressor that belongs to group G of the SOX gene family. Sox-15 is widely expressed in fetal and adult tissues, with highest level found in fetal spinal cord, adult brain and adult testis. Recently, SOX-15 has been identified as having transcriptional control in mouse embryonic stem cells pluripotency along with SOX-2. SOX-15 is also expressed in myoblasts and regulates skeletal muscle regeneration and differentiation.

Immunogen

GST-tagged recombinant protein corresponding to human SOX-15.

Application

Anti-SOX-15 Antibody, clone 1A2.1 is an antibody against SOX-15 for use in western blotting & ICC.
Immunocytochemsitry Analysis: A 1:50 dilution of this antibody detected SOX-15 in a undifferentiated H9 human embryonic stem cells.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Quality

Evaluated by Western Blotting in a undifferentiated H9 human embryonic stem cell lysate. Western Blotting Analysis: 1.0 µg/mL of this antibody detected SOX-15 in a undifferentiated H9 human embryonic stem cells.

Target description

~ 36 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service