Skip to Content
Merck
All Photos(2)

Key Documents

MABD89

Sigma-Aldrich

Anti-Podocalyxin-like protein I (Cytotoxic) Antibody, clone mAb 84

clone mab 84, from mouse

Synonym(s):

Podocalyxin-like protein I, GCTM-2 antigen, Gp200

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€446.00

€446.00


Estimated to ship on26 April 2025



Select a Size

Change View
100 μG
€446.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€446.00


Estimated to ship on26 April 2025


biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

mab 84, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PODXL(5420)

General description

Podocalyxin-like protein 1 (PODXL) is a transmembrane protein known to be heavily sialyated and sulfated that is localized on the apical surface of podocytes and on vascular endothelia. Podocalyxin protein is expressed on the surface of undifferentiated human embryonic stem cells. Originally, the protein was characterized as a marker for vasculature and podocytes, but expression has since been observed in platelets, hematopoietic precursor cells and mesothelial cells from the coelmic cavity lining. Studies have outlined two distinct and seemingly adverse roles for this protein; it acts as an anti-adhesion molecule in renal glomerulae podocytes, but functions as an adhesion molecule within vasculature on high endothelial venules. Podocalyxin expression has been implicated in the development of aggressive cancers, beta catenin signaling, angiogenisis, and both myeloid and acute lymphoblastic leukemias.

Specificity

This antibody recognizes the extracellular domain of Podocalyxin-like protein 1.
This antibody has been shown to select against undifferentiated human embryonic stem cells (hESC) and to be functionally cytotoxic against these undifferentiated hESC populations (Choo, A., et al., 2008).

Immunogen

HES-3 Human embryonic stem cell line

Application

Anti-Podocalyxin-like protein I (Cytotoxic) Antibody, clone mAb 84 is an antibody against Podocalyxin-like protein I (Cytotoxic) for use in Western Blotting, ICC, Flow Cytometry.
This antibody was shown to induce cell death of undifferentiated, but not differentiated hESC, within 30 minutes of incubation (Choo, A., et al. (2008). Stem Cells 26:1454-1463). This antibody has been shown to work in Western Blot applications (Choo, A., et al. (2008). Stem Cells, 26:1454–1463).
This antibody has been shown to work in Flow Cytometry applications (Choo, AB., et al. (2008). Stem Cells, 26:1454–1463).

Quality

Evaluated by Immunocytochemistry in H9 human embryonic stem cells. Immunocytochemistry Analysis: A 1:500 dilution of this antibody detected Podocalyxin-like protein I in H9 human embryonic stem cells.

Target description

55 kDa calculated

Linkage

Replaces: MAB4414

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Kang et al.
Stem cells and development, 25(7), 556-568 (2016-02-18)
Podocalyxin-like protein (PODXL) is a member of CD34 family proteins. It is the protein that carries many post-translational epitopes responsible for various pluripotent surface markers including TRA-1-60, TRA-1-81, GCTM2, GP200, and mAb84. However, PODXL has not attracted the attention of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service