Skip to Content
Merck
All Photos(1)

Key Documents

MABD86

Sigma-Aldrich

Anti-CD349/Frizzled 9 Antibody, clone W3C4

clone W3C4-hMSC, from mouse

Synonym(s):

CD349, Frizzled-9, Frizzled 9, FZ-9, FzE6, FzD3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€370.00

€370.00


Estimated to ship on08 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€370.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€370.00


Estimated to ship on08 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

W3C4-hMSC, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FZD9(8326)

Related Categories

General description

CD349 is a member of the Frizzled family, known as Frizzled-9 (Fz9, FZD9, hFz9, or FzE6) which was originally described as FZD3. Ten frizzled member have been identified in humans. FZD9 is a seven transmembrane G-protein-coupled receptor (GPCR), a receptor for Wnt proteins. It is involved in the activation of Wnt/β-catenin pathway (canonic Wnt signaling). FZD9 is predominately expressed in fetal and adult brain, testis, eye, skeletal muscle and kidney and is also expressed on Mesenchymal stem cells, and neural precursor cells. Frizzled proteins play important roles in controlling cell fate, proliferation, migration, tissue architecture and organogenesis during embryonic development and in adult organisms. FZD9 is an important factor for nervous system development and is critical for hippocampal development.

Immunogen

Whole WERI-RB-1 cells

Application

Anti-CD349/Frizzled 9 Antibody, clone W3C4 is an antibody against CD349/Frizzled 9 for use in Flow Cytometry.
Research Category
Stem Cell Research

Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Developmental Signaling

Quality

Evaluated by Flow Cytometry on WERI-RB-1 cells Flow Cytometry Analysis: 4 µg/mL of this antibody detected CD349/Frizzled 9 on 1X10^5 WERI-RB-1 cells.

Target description

65 kDa calculated

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
WERI-RB-1 cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service