Skip to Content
Merck
All Photos(1)

Key Documents

MABD34

Sigma-Aldrich

Anti-Werner′s syndrome helicase (WRN) Antibody, clone 20A11.1

clone 20A11.1, from mouse

Synonym(s):

Werner syndrome ATP-dependent helicase, DNA helicase, RecQ-like type 3, RecQ3, Exonuclease WRN, RecQ protein-like 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€487.00

€487.00


Estimated to ship on14 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€487.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€487.00


Estimated to ship on14 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

20A11.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WRN(7486)

General description

The Werner’s syndrome helicase (RecQ-like type 3; RecQ3; Exonuclease WRN) is a ubiquitously expressed nuclear protein belonging to the RecQ family. It functions as a helicase and exonuclease enzyme, and plays a crucial role in the formation of DNA replication centers. This protein is also involved in DNA repair and recombination, and may be involved in transcriptional events mediated by RNA polymerase II. It is regulated by phosphorylation and is targeted by the DNA-dependent protein kinase enzyme. Disrupted expression of the gene encoding the Werner’s syndrome helicase protein often results in the production of truncated Werner’s syndrome helicase proteins. Over 50 gene mutations may contribute to this effect—and lead to the development of Werner syndrome, a condition characterized by accelerated aging and early onset of age-related conditions such as cancer, atherosclerosis, and osteoporosis.

Immunogen

GST-tagged recombinant protein corresponding to human WRN.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
Use Anti-WRN Antibody, clone 20A11.1 (Mouse Monoclonal Antibody) validated in WB to detect WRN clone 20A11.1 also known as Werner syndrome ATP-dependent helicase, RecQ3, Exonuclease WRN.

Quality

Evaluated by Western Blot in HEK293 cell lysate.

Western Blot Analysis: A 1:2,000 dilution of this antibody detected Werner′s syndrome helicase (WRN) in 10 µg of HEK293 cell lysate.

Target description

~162 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephane Ferretti et al.
Nature, 629(8011), 443-449 (2024-04-25)
The Werner syndrome RecQ helicase WRN was identified as a synthetic lethal target in cancer cells with microsatellite instability (MSI) by several genetic screens1-6. Despite advances in treatment with immune checkpoint inhibitors7-10, there is an unmet need in the treatment

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service