Skip to Content
Merck
All Photos(1)

Key Documents

MABC683

Sigma-Aldrich

Anti-SMAD6 Antibody, clone 5H3

ascites fluid, clone 5H3, from mouse

Synonym(s):

Mothers against decapentaplegic homolog 6, MAD homolog 6, Mothers against DPP homolog 6, SMAD family member 6, SMAD 6, Smad6, hSMAD6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€397.00

€397.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€397.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€397.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

5H3, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SMAD6(4091)

General description

SMAD6 is the gene name for a protein that is a homolog of the fly gene Mothers against decapentaplegic, and in humans it is called Mothers against decapentaplegic homolog 6 (MAD homolog 6). Other names include SMAD family member 6 and Mothers against DPP homolog 6 and another gene name of MADH6. By whatever name, the protein is a transcription factor that binds to regulatory elements within the nucleus and mediates transcription. The effect is to act as a mediator of TGF-beta and BMP mediated anti-inflammatory signaling activity. SMAD6 suppresses IL1R-TLR signaling, thus preventing NF-kappa B activation and activation of proinflammatory genes. SMAD6 interacts with numerous other intracellular proteins including NEDD4L, STAMBP, PRKK, RNF111, and AXIN1. SMAD6 is expressed in a wide variety of tissues with higher levels being seen in lung and cardiac tissues. Recently SMAD6 has been shown to promote neuronal differentiation by inhibiting the Wnt/beta catenin signaling pathway in addition to inhibiting the BMP signaling. Finally in cancer SMAD6 via its suppression of TGF-beta induced growth inhibition can potentiate the transformed phenotype. Moreover SMAD6 is often over expressed in many cancers, which suppresses G0-G1 arrest and apoptosis which might otherwise normally occur, thus leaving SMAD6 playing a critical role in supporting cancer cell growth and survival. EMD-Millipore’s Anti-SMAD6 mouse monoclonal antibody has been tested in western blot against human A431, HeLa, and Jurkat cell lysates.

Immunogen

Purified recombinant fragment of human SMAD6 expressed in E. Coli.

Application

Detect Smad using this mouse monoclonal antibody, Anti-SMAD6 Antibody & clone 5H3 validated for use in western blotting.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blot in A431, HeLa, and Jurkat cell lysates.

Western Blot Analysis: A 1:500-2000 dilution of this antibody detected SMAD6 in A431, HeLa, and Jurkat cell lysates.

Optimal working dilutions must be determined by end user.

Target description

~55 kDa observed

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
A431, HeLa, and Jurkat cell lysates

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service