Skip to Content
Merck

MABC606

Anti-RNF43 Antibody, clone 11C3.1

clone 11C3.1, from mouse

Synonym(s):

E3 ubiquitin-protein ligase RNF43, RING finger protein 43

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
11C3.1, monoclonal
Application:
WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

11C3.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... RNF43(54894)

General description

E3 ubiquitin-protein ligase RNF43 (UniProt Q68DV7; also known as RING finger protein 43) is encoded by the RNF43 gene (Gene ID 54894) in human. RNF43 and the homologous ZNRF3 are cell-surface transmembrane E3 ubiquitin ligases upregulated upon Wnt/ -catenin signaling and act as potent Wnt signaling pathway negative feedback regulators by mediating ubiquitination of Frizzled (FZD) and LRP6 for lysosomal degradation. RNF43/ZNRF3 activity is counteracted by stem cell growth factor R-spondin that induces membrane ZNRF3/RNF43 clearance by binding ZNRF3/RNF43 and LGR4/LGR5. Physical interaction with Dishevelled (DVL) is essential for ZNRF3/RNF43 Wnt inhibitory activity, and DVL knockdown significantly increases cell surface levels of FZD and LRP6. In addition, RNF43 suppresses canonical and noncanonical Wnt signaling via distinct mechanisms. While interaction between the extracellular protease-associated (PA) domain and the cysteine-rich domain (CRD) of FZD and RNF43 intracellular RING finger domain is necessary for canonical Wnt/ -catenin signaling inhibition, interaction between FZD PDZ domain and RNF43 C-terminal cytoplasmic region is essential for suppressing noncanonical Wnt signaling. R-spondin overexpression through gene translocation and loss-of-function RNF43/ZNRF3 gene mutations are found in human cancers as means to escape from Wnt signaling negative feedback regulation. Human RNF43 is produced with a signal peptide (a.a. 1-23) sequence, which is posttranslationally removed to yiled the mature enzyme with an N-terminal extracellular domain (a.a. 24-197), a transmembrane domain (a.a. 198-218), and a large cytoplasmic region (a.a. 219-783) that contains an atypical RING-type zinc finger (a.a. 272-313) and a pro-rich (a.a. 569-760) region.
~95 kDa observed. Target band size appears larger than the calculated molecular weights of 83.20/85.72 kDa (isoform 1 mature/pro-form), 78.71/81.23 kDa (isoform 2 mature/pro-form), 72.11 kDa (isoform 3) 92.52/95.04 kDa (isoform 4 mature/pro-form) due to glycosylation. Uncharacterized bands may be observed in some lysate(s).

Immunogen

GST-tagged recombinant human RNF43 fragment from the cytoplasmic pro-rich region.

Application

Detect RNF43 using this mouse monoclonal Anti-RNF43, clone 11C3.1 Antibody, Cat. No. MABC606, validated for use in Western Blotting.
Research Category
Apoptosis & Cancer

Biochem/physiol Actions

Clone 11C3.1 epitope lies within the cytoplasmic pro-rich region present in all four spliced isoforms of human RNF43 reported by UniProt (Q68DV7).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in HCT116 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected RNF43 in 10 µg of HCT116 human colon carcinoma cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library