Skip to Content
Merck

MABC593

Anti-Astrin/SPAG5, clone 8B2 Antibody

clone 8B2, from rat

Synonym(s):

Sperm-associated antigen 5, Astrin, Deepest, Mitotic spindle-associated protein p126, MAP126

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
8B2, monoclonal
Application:
ICC, IP, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

8B2, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG2cκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SPAG5(10615)

General description

134 kDa calculated
Astrin/SPAG5 is a microtubule binding and cytoskeleton associated signaling protein that plays not only a critical role in mitotic spindle and chromosome segregation but also as an important signal transducer in mTOR and GSK3 beta signaling pathways. Astrin/SPAG5 is localized in the cytoplasm and/or mitotic apparatus (in dividing cells) and centrosomes. In non-dividing cells Astrin/SPAG5 is associated with microtubules unless under cellular stress, then it is localized in stress granules. Astrin/SPAG5 is expressed in many tissues but is very high in testis and sperm and is regulated via the cell cycle highest levels are seen in prophase. Recent research has focused on Astrin/SPAG5 critical role in regulating mTORC1 and its effects on growth and survival. When cells are stress Astrin/SPAG5 inhibits mTORC1 association and recruits the mTORC1 component raptor to stress granules (SGs), thereby preventing mTORC1-hyperactivation-induced apoptosis. Because cancerous tumor cells are often stressed, this mechanism is believed to be key to their survival, thus Astrin/SPAG5 activity repression is being examined as potential anti-cancer therapy.

Immunogen

Linear peptide corresponding to human Astrin/SPAG5.

Application

Immunocytochemistry Analysis: A representative lot detected Astrin/SPAG5 HeLa cells (Thedieck, K., et al. (2013). Cell. 154:859-874).
Western Blotting Analysis: A representative lot detected Astrin/SPAG5 in SiAstrin (Dr. Elisabeth Kremmer).
Immunoprecipitation Analysis: A representative lot immunoprecipitated Astrin/SPAG5 in SiAstrin (Dr. Elisabeth Kremmer).
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-Astrin/SPAG5, clone 8B2 Antibody is validated for use in Immunocytochemistry and Western Blotting and Immunoprecipitation for the detection of Astrin/SPAG5.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2cκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Immunocytochemistry in HeLa and A431 cells.

Immunocytochemistry Analysis: A 1:50 dilution of this antibody detected Astrin/SPAG5 in HeLa and A431 cells.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library