Skip to Content
Merck
All Photos(2)

Key Documents

MABC557

Sigma-Aldrich

Anti-FANCA Antibody, clone 5G9

clone 5G9, from mouse

Synonym(s):

Fanconi anemia group A protein, Protein FACA

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€475.00

€475.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€475.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€475.00


Estimated to ship on19 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5G9, monoclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FANCA(2175)

General description

FANCA, also named as FAA, FACA and FANCH, belongs to the multisubunit FA complex which is not found in patients with Fanconi anemia. FANCA may be involved in interstrand DNA cross-link repair and in the maintenance of normal chromosome stability. Mutations in FANCA have been associated with Fanconi anemia (FA) which is a genetically heterogeneous, autosomal recessive disorder characterized by progressive pancytopenia, a diverse assortment of congenital malformations, and a predisposition to the development of malignancies.

Immunogen

KLH-conjugated linear peptide corresponding to the C-terminus of human FANCA.

Application

Anti-FANCA Antibody, clone 5G9 is a highly specific mouse monoclonal antibody, that targets FANCA & has been tested in western blotting & IP.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected FANCA in 10 µg of HeLa cell lysate.

Western Blotting Analysis: A representative lot from an independent laboratory detected FANCA in normal HSC93 and FA mutant lymphoblast whole cell extracts. The same lot also detected FANCA in HeLa whole cell extracts and in FANCA transfected PD430 whole cell extracts (Tremblay, C. S., et al. (2008). Blood. 112(5):2062-2070.).

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated FANCA from yeast extracts expressing FANCA. The same lot also immunoprecipiated FANCA from 293T whole cell extracts expressing FANCA (Tremblay, C. S., et al. (2008). Blood. 112(5):2062-2070.)

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated FANCA from FANCA wild type FA lymphoblast cell lysate (de Winter, J. P., et al. (2000). Hum Mol Genet. 9(18):2665-2674.).

Quality

Evaluated by Western Blotting in Raji cell lysates.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected FANCA in 10 µg of Raji cell lysate.

Target description

~165 kDa observed

Physical form

Format: Purified

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anudari Munkhjargal et al.
International journal of molecular sciences, 22(15) (2021-08-08)
Promyelocytic leukemia (PML) protein is the core component of subnuclear structures called PML nuclear bodies that are known to play important roles in cell survival, DNA damage responses, and DNA repair. Fanconi anemia (FA) proteins are required for repairing interstrand
Mafei Xu et al.
Science advances, 5(10), eaax6366-eaax6366 (2019-10-22)
Alternative lengthening of telomeres (ALT) is known to use homologous recombination (HR) to replicate telomeric DNA in a telomerase-independent manner. However, the detailed process remains largely undefined. It was reported that nuclear receptors COUP-TFII and TR4 are recruited to the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service