Skip to Content
Merck
All Photos(1)

Key Documents

MABC43

Sigma-Aldrich

Anti-Mcl-1 Antibody, clone RC13

clone RC13, from mouse

Synonym(s):

myeloid cell leukemia sequence 1 (BCL2-related), myeloid cell leukemia sequence 1, isoform 1, Bcl-2-like protein 3, Bcl-2-related protein EAT/mcl1, myeloid cell leukemia ES, induced myeloid leukemia cell differentiation protein Mcl-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RC13, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MCL1(4170)

General description

Mcl-1 (Induced myeloid leukemia cell differentiation protein) is a potential transmembrane protein associated with the mitochondria belonging to the Bcl-2 family of proteins. Members of the Bcl-2 family have known participation in regulatory roles for apoptosis, cell growth, and differentiation. The hydrophobic carboxyl terminus is conserved among members of this family and in other members has proven to be a transmembrane region. Mcl-1 is found to be expressed early in myeloid differentiation. Two separate isoforms of Mcl-1 exist with opposing functions. Full length Mcl-1 (Isoform 1) promotes cell viability, over proliferation. Isoform 2 results from the skipping of the central exon of Mcl-1 during mRNA processing and the joining of the first and third exons. The function of the resulting protein induces apoptosis when accumulated. Pro-apoptotic function is shown to increase when isoform 1 and 2 dimerize.

Immunogen

Recombinant protein corresponding to human Mcl-1.

Application

Detect Mcl-1 using this Anti-Mcl-1 Antibody, clone RC13 validated for use in WB, IH & IP.
Immunohistochemistry Analysis: A previous lot was used by an independent laboratory in follicular lymphoma, diffuse large cell lymphoma, and polymorphic lymphoma tissues. (Zhou, P., et al. (2001). Blood. 97(12):3902-3909).

Immunoprecipitation Analysis: A previous lot was used by an independent laboratory in IP (Han, J., et al. (2005). Biological Chemistry. 280:16383-16392).
Research Category
Apoptosis & Cancer
Research Sub Category
BCL2 & Inhibition

Quality

Evaluated by Western Blot in Raji cell lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected Mcl-1 in 10 µg of Raji cell lysate.

Target description

~37 kDa observed

Linkage

Replaces: MAB4602

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Raji cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qi Shen et al.
Cell death & disease, 9(10), 986-986 (2018-09-27)
Lung cancer is the leading cause of death in the United States, with non-small cell lung cancers (NSCLC) accounting for 85% of all cases. By analyzing the expression profile of the pro-apoptotic and anti-apoptotic proteins, we have assigned NSCLCs into
Lindy Vernooij et al.
Molecular cancer therapeutics, 20(6), 1161-1172 (2021-04-15)
Neuroblastoma tumors frequently overexpress the anti-apoptotic protein B-cell lymphoma/leukemia 2 (BCL-2). We previously showed that treating BCL-2-dependent neuroblastoma cells with the BCL-2 inhibitor venetoclax results in apoptosis, but unfortunately partial therapy resistance is observed. The current study describes the identification
Raju R Rayavarapu et al.
The Journal of biological chemistry, 290(14), 8722-8733 (2015-02-15)
The metastasis of cancer cells from the site of the primary tumor to distant sites in the body represents the most deadly manifestation of cancer. In order for metastasis to occur, cancer cells need to evade anoikis, which is defined
Adriana E Tron et al.
Nature communications, 9(1), 5341-5341 (2018-12-19)
Mcl-1 is a member of the Bcl-2 family of proteins that promotes cell survival by preventing induction of apoptosis in many cancers. High expression of Mcl-1 causes tumorigenesis and resistance to anticancer therapies highlighting the potential of Mcl-1 inhibitors as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service