Skip to Content
Merck
All Photos(1)

Key Documents

MABC282

Sigma-Aldrich

Anti-BAG3 Antibody, clone AC-1

clone AC-1, from mouse

Synonym(s):

BAG family molecular chaperone regulator 3, BAG-3, Bcl-2-associated athanogene 3, Bcl-2-binding protein Bis, Docking protein CAIR-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€327.00

€327.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€327.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€327.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

AC-1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BAG3(9531)

General description

Bcl2-associated athanogene 3 (BAG3), also known as CAIR-1 or Bis, belongs to a family of six evolutionarily conserved co-chaperone proteins that contain a characteristic C-terminal BAG domain. In most cell types, BAG3 is localized to the cytoplasm, where it regulates the heat shock protein 70 chaperone. BAG3 is activated in response to stressful stimuli, and plays a critical role in modulating multiple cellular processes, including apoptosis, autophagy, embryonic development, and actin rearrangement. Abnormal expression of BAG3 results cardiomyopathy dilated type 1HH.

Application

Detect BAG3 using this Anti-BAG3 Antibody, clone AC-1 validated for use in Immunoprecipitation, IHC(P).

Quality

Evaluated by Immunoprecipitation in HeLa cell lysate.

Immunoprecipitation Analysis: 10 µg of this antibody immunoprecipitated BAG3 from HeLa cell lysate.

Target description

~62 kDa observed

Physical form

Format: Purified

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Becky Lou et al.
Journal of oncology, 2012, 491685-491685 (2012-06-22)
We performed comparative global proteomics analyses of patient-matched primary (686Tu) and metastatic (686Ln) OSCC cells. The metastatic OSCC 686Ln cells showed greater in vitro migratory/invasive potential and distinct cell shape from their parental primary 686Tu cells. Ettan DIGE analysis revealed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service