Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μg | Check Cart for Availability | € 420,00 |
About This Item
biological source
mouse
Quality Segment
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
6C10.1, monoclonal
species reactivity
human
technique(s)
immunohistochemistry: suitable (paraffin), western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... RAB27B(5874)
General description
Immunogen
Application
Apoptosis & Cancer
Apoptosis - Additional
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: 1.0 µg/mL of this antibody detected Rab27B in 10 µg of human prostate tissue lysate.
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| species reactivity human | species reactivity human, rat, mouse | species reactivity human | species reactivity human |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| clone 6C10.1, monoclonal | clone polyclonal | clone 5G8.1, monoclonal | clone polyclonal |
| antibody form purified immunoglobulin | antibody form serum | antibody form purified immunoglobulin | antibody form affinity isolated antibody |
| Gene Information human ... RAB27B(5874) | Gene Information human ... RAB27B(5874) | Gene Information human ... RAB7A(7879) | Gene Information human ... RAB37(326624) |
| biological source mouse | biological source rabbit | biological source mouse | biological source rabbit |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



