Skip to Content
Merck
All Photos(1)

Key Documents

MABC1723

Sigma-Aldrich

Anti-TM4SF1 Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€152.00
100 μL
€383.00

€152.00


Estimated to ship on09 April 2025


Request a Bulk Order

Select a Size

Change View
25 μL
€152.00
100 μL
€383.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€152.00


Estimated to ship on09 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

8G4, monoclonal

mol wt

calculated mol wt 21.63 kDa

packaging

antibody small pack of 100 μL

isotype

IgG1κ

UniProt accession no.

shipped in

dry ice

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Gene Information

human ... TM4SF1(4071)

General description

Transmembrane 4 L6 family member 1 (UniProt: P30408; also known as Membrane component chromosome 3 surface marker 1, Tumor-associated antigen L6, TM4SF1) is encoded by the TM4SF1 (also known as M3S1, TAAL6) gene (Gene ID: 4071) in human. TM4SF1 is a multi-pass membrane protein of the L6 tetraspanin family with tetraspanin topology. It has three cytoplasmic domains, four transmembrane domains, and two extracellular domains. It is highly expressed in lung, breast, colon, and ovarian carcinoma. It is highly and selectively expressed on the plasma membranes of tumor cells and on tumor-associated endothelium. It regulates cell motility and proliferation. In PANC-1 and AsPC-1 cells, TM4SF1 is reported to mediate the expression and activities of matrix metallopeptidase 2 (MMP2) and matrix metallopeptidase 9 (MMP9), which are considered as the major regulators of extracellular matrix degradation to increase the cells migration and invasion. Clone 8G4 is reported to be highly effective in reducing blood vessel formation in Matrigel plugs and is shown to destroy PC3 cells and vascular components incorporated into the Matrigel mixture. (Ref.: Yang, J., et al. (2017). Sci. Rep. 7; Article 45895; Sciuto, TE., et al. (2015). Biochem. Biophys. Res. Commun. 465(3): 338 343; Lin, C., et al. (2014). Angiogenesis. 17(4); 897-907).

Specificity

Clone 8G4 is a mouse monoclonal antibody that detects human Transmembrane 4 L6 family member 1 (TM4SF1). It targets an epitope within the extracellular loop 2.

Immunogen

Human umbilical vein endothelial cells (HUVEC) overexpressing human TM4SF1.

Application

Quality Control Testing

Evaluated by Immunocytochemistry in human umbilical vein endothelial cells (HUVEC).

Immunocytochemistry Analysis: A 1:250 dilution of this antibody detected TM4SF1 in human umbilical vein endothelial cells (HUVEC).

Tested Applications

Flow Cytometry Analysis: A representative lot detected TM4SF1 in Flow Cytometry applications (Lin, C.I., et. al. (2014). Angiogenesis. 17(4):897-907).

Western Blotting Analysis: A representative lot detected TM4SF1 in Western Blotting applications (Sciuto, T.E., et. al. (2015). Biochem Biophy Res Commun. 465(3):338-443).

Immunoprecipitation Analysis: A representative lot immunoprecipitated TM4SF1 in Immunoprecipitation applications (Sciuto, T.E., et. al. (2015). Biochem Biophy Res Commun. 465(3):338-443).

Immunocytochemistry Analysis: A representative lot detected TM4SF1 in Immunocytochemistry applications (Lin, C.I., et. al. (2014). Angiogenesis. 17(4):897-907).

Electron Microscopy: A representative lot detected TM4SF1 in Electron Microscopy applications (Sciuto, T.E., et. al. (2015). Biochem Biophy Res Commun. 465(3):338-443).

Inhibition Analysis: A representative lot blocked the formation new blood vessels in Matrigel plugs. (Lin, C.I., et. al. (2014). Angiogenesis. 17(4):897-907).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
This mouse monoclonal Anti-TM4SF1, clone 8G4 , Cat. No. MABC1723 is tested for use in Immunocytochemistry, Inhibition, Electron Microscopy, Immunoprecipitation, Western Blotting and Flow Cytometry, for the detection of Transmembrane 4 L6 family member 1.

Physical form

Purified mouse monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service