Skip to Content
Merck
All Photos(1)

Key Documents

MABC1595

Sigma-Aldrich

Anti-RIPK3/RIP3 Antibody, clone 8G7

Synonym(s):

Anti-RIPK3 Antibody, Clone 8G7 Anti-RIPK3, RIPK3 Detection Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€153.00
100 μL
€383.00

€153.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€153.00
100 μL
€383.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€153.00


Check Cart for Availability

Request a Bulk Order

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

8G7, monoclonal

mol wt

calculated mol wt 53.32 kDa
observed mol wt ~53 kDa

species reactivity

mouse

packaging

antibody small pack of 100 μL

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

dry ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Receptor-interacting serine/threonine-protein kinase 3 (UniProt: EC:2.7.11.1, RIP-like protein kinase 3, Receptor-interacting protein 3, RIP-3, mRIP3) is encoded by the Ripk3 (also known as Rip3) gene (Gene ID: 56532) in murine species. RIPK3 is a serine/threonine protein kinase that is expressed in embryo and adult spleen, liver, testis, brain, and lung. Its protein kinase domain is localized in amino acids 22 to 293 in the N-terminal half and it also has a C-terminal RIP homolytic interaction motif (RHIM; aa 440-461), which mediates interaction with the RHIM motif of RIPK1 and facilitate the formation of a necrosome. RIPK3 is shown to activate both necroptosis and apoptosis. Activated RIPK3 forms a necrosis-inducing complex and mediates phosphorylation of MLKL, promoting MLKL localization to the plasma membrane and execution of programmed necrosis that is characterized by calcium influx and plasma membrane damage. Its location is mainly cytoplasmic, however, following infection, it can also be detected in the nucleus. RIPK3 also regulates apoptosis that is dependent on RIPK1, FADD, and CASP8, and is independent of MLKL and RIPK3 kinase activity. It phosphorylates RIPK1 and then RIPK1 and RIPK3 undergo reciprocal auto- and trans-phosphorylation. In some cell types, RIPK3 is also reported to restrict viral replication by promoting cell death-independent responses. Clone 8G7 is raised against an extended kinase domain (residues 2-353) of mouse RIPK3 and can detect forms that may not be detected by antibodies directed towards the C-terminus of the full-length RIPK3. (Ref.: Samson, AL., et al. (2021). Cell Death Differ. https://doi.org/10.1038/s41418-021-00742-x; Petrie, EJ., et al. (2019). Cell Rep. 28(13); 3309-3319).

Specificity

Clone 8G7 is a rat monoclonal antibody that detects murine Receptor-interacting serine/threonine-protein kinase 3 (RIPK3). It targets an epitope within the extended kinase domain that includes most of the N-terminal half and a part of C-terminal half.

Immunogen

Recombinant fragment corresponding to 352 amino acids from the extended kinase domain of murine Receptor-interacting serine/threonine-protein kinase 3 (RIPK3).

Application

Quality Control Testing

Evaluated by Western Blotting in Mouse dermal fibroblast lysates.

Western Blotting Analysis: A 1:500 dilution of this antibody detected RIPK3/RIP3 in Mouse dermal fibroblast lysates.

Tested Applications

Western Blotting Analysis: A representative lot detected RIPK3/RIP3 in Western Blotting applications (Petrie, E.J., et. al. (2019). Cell Rep. 28(13):3309-3319).

Immunofluorescence Analysis: A representative lot detected RIPK3/RIP3 in Immunofluorescence applications (Samson, A.L., et. al. (2021). Cell Death Differ. doi: 10.1038/s41418-021-00742-x)

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-RIPK3/RIP3, clone 8G7, Cat. No. MABC1595, is a rat monoclonal antibody that detects RIPK3/RIP3 and is tested for use in Immunofluorescence and Western Blotting.

Physical form

Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service