Skip to Content
Merck
All Photos(1)

Key Documents

MABC1120

Sigma-Aldrich

Anti-PD-L2 Antibody, clone 366C.9E5

clone 366C.12.9E5.E10.19.1F8, from mouse

Synonym(s):

Programmed cell death 1 ligand 2, PD-1 ligand 2, PDCD1 ligand 2, Programmed death ligand 2, Butyrophilin B7-DC, B7-DC, CD273

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€154.00
100 μG
€403.00

€154.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€154.00
100 μG
€403.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€154.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

366C.12.9E5.E10.19.1F8, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

General description

Programmed cell death 1 ligand 2 (UniProt: Q9BQ51; also known as PD-1 ligand 2, PD-L2, PDCD1 ligand 2, Programmed death ligand 2, Butyrophilin B7-DC, B7-DC, CD273) is encoded by the PDCD1LG2 (also known as B7DC, CD273, PDCD1L2, PDL2) gene (Gene ID: 80380) in human. Three isoforms of PD-L2 have been reported that are produced by alternative splicing. Isoforms 1 and 2 are single-pass type I membrane proteins and isoform 3 is a secreted protein. PD-1 and PD-1 ligands 1 and 2 (PD-L1 and PD-L2) are B7:CD28 family members that regulate T cell activation and peripheral tolerance. When engaged together with the TCR, the interaction of PD-1 with its ligands delivers an inhibitory signal to T cell proliferation and cytokine production. While PD-L1 is broadly expressed in hematopoietic and non-hematopoietic cells, PD-L2 expression is highly restricted to antigen presenting cells (APCs), including dendritic cells (DCs) and macrophages. The PD-1 pathway plays a key role in the progressive loss of effector T cell responses during chronic HIV infection. Under some conditions, blockade of this pathway can restore many T cell functions. PD-L2 is initially produced with signal peptide (aa 1-19) that is removed to produce the mature protein that contains a large extracellular region (aa. 20-220), a transmembrane domain (aa 221-241), and a cytoplasmic tail (aa 242-273).

Specificity

Clone 366C.12.9E5.E10.19.1F8 specifically detects PD-L2 in human cells. It targets an epitope within the extracellular domain.

Immunogen

Epitope: extracellular domain
Recombinant protein fragment corresponding to the extracellular domain of human PD-L2 (as an Ig fusion protein).

Application

Anti-PD-L2, clone 366C.9E5, Cat. No. MABC1120, is a mouse monoclonal antibody that detects Programmed cell death 1 ligand 2 (PD-L2) and has has been tested for use in Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected PD-L2 in human prostate tissue.

Immunohistochemistry Analysis: A representative lot detected PD-L2 in Immunohistochemistry applications (Sridharan, V., et. al. (2016). Cancer Immunol Res. 4(8):679-87; Roemer, M.G., et. al. (2016). J Clin Oncol. 34(23):2690-7; Chong, L.C., et. al. (2016). Blood. 128(9):1206-13; Calles, A., et. al. (2015). J Thorac Oncol. 10(12):1726-35).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Immunohistochemistry (Paraffin) in human tonsil tissue.

Immunohistochemistry Analysis: A 1:250 dilution of this antibody detected PD1-L2 in human tonsil tissue.

Target description

30.96 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selim Chaib et al.
Nature cancer, 5(3), 448-462 (2024-01-25)
Chemotherapy often generates intratumoral senescent cancer cells that strongly modify the tumor microenvironment, favoring immunosuppression and tumor growth. We discovered, through an unbiased proteomics screen, that the immune checkpoint inhibitor programmed cell death 1 ligand 2 (PD-L2) is highly upregulated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service