Skip to Content
Merck
All Photos(1)

Key Documents

MAB880-K

Sigma-Aldrich

Anti-HIV Antibody, p24, clone 7A8.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

antibody form

ascites fluid

Quality Level

clone

7A8.1
monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

Physical form

Core protein p24. No cross-reactivity observed to other HIV proteins.

Storage and Stability

Maintain at -20°C Avoid multiple freeze-thaw cycles.

Other Notes

p24 isolated from human (native) HIV infected cells.

Disclaimer

This product is intended for use by IVD manufacturers.
Contact customer service for assistance: [email protected]
Minimum order quantity required to qualify. Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service