Skip to Content
Merck
All Photos(1)

Key Documents

MAB8685

Sigma-Aldrich

Anti-Herpes Simplex Virus II gB Antibody, clone 1-G-10

clone 1-G-10, Chemicon®, from mouse

Synonym(s):

HSVII gB

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€370.00

€370.00


Estimated to ship on28 March 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€370.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
1-G-10, monoclonal
application:
ELISA
IF
WB
species reactivity:
human
technique(s):
ELISA: suitable
immunofluorescence: suitable
western blot: suitable
citations:

€370.00


Estimated to ship on28 March 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

1-G-10, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

Specificity

HSV (II) gB antigen

Immunogen

Produced against an HSV (II) infected cell.

Application

Anti-Herpes Simplex Virus II gB Antibody, clone 1-G-10 detects level of Herpes Simplex Virus II gB & has been published & validated for use in ELISA, IF & WB.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
Western Blot: 1:1000

ELISA: 1:10,000

Immunofluorescence: 1:1000

Optimal working dilutions must be determined by end user.

Linkage

Replaces: MAB8571

Physical form

Format: Purified
Protein A purified immunoglobulin - Ig fraction. Liquid in phosphate buffered saline, pH 7.2 with 0.01% sodium azide.

Storage and Stability

Store at -20°C in undiluted aliquots. May be stored at 2-8°C for short term use. Avoid repeated freeze-thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service