Skip to Content
Merck
All Photos(1)

Key Documents

MAB8594

Sigma-Aldrich

Anti-RSV Antibody, fusion protein, clone 143-6C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

antibody form

purified antibody

Quality Level

clone

143-6C
monoclonal

species reactivity

mouse, human

concentration

(Please refer to the Certificate of Analysis for the lot-specific concentration.)

technique(s)

immunofluorescence: suitable

isotype

IgG

General description

V1680E

Specificity

Reacts with RSV fusion protein.

Application

Indirect immunofluorescence.

Final working dilutions must be determined by end user.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service