Skip to Content
Merck
All Photos(1)

Key Documents

MAB8251

Sigma-Aldrich

Anti-Influenza A Antibody, nucleoprotein, clones A1, A3 Blend

ascites fluid, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€605.00

€605.00


Estimated to ship on29 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZMS1020

Request a Bulk Order

Select a Size

Change View
100 μL
€605.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€605.00


Estimated to ship on29 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZMS1020

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

A1, monoclonal
A3, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG1 (A3)
IgG2a (A1)

shipped in

wet ice

Specificity

All subtypes of Influenza A (H1N1, H2N2 and H3N2 and H5N1). Specific for the nucleoprotein antigen. No cross-reactivity seen to Influenza B, Parainfluenza 1, 2 or Adenovirus or RSV.

Immunogen

Influenza A blend.

Application

Indirect immunofluorescence at 1:100-1:1000.

Optimal working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
This Anti-Influenza A Antibody, nucleoprotein, clones A1, A3 Blend is validated for use in IF for the detection of Influenza A.

Physical form

Ascites fluid containing no preservatives.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Influenza Control Slides, Catalogue Number 5010-5

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nara Lee et al.
Nucleic acids research, 45(15), 8968-8977 (2017-09-16)
Influenza A virus (IAV) genomes are composed of eight single-stranded RNA segments that are coated by viral nucleoprotein (NP) molecules. Classically, the interaction between NP and viral RNA (vRNA) is depicted as a uniform pattern of 'beads on a string'.
Rui Jia et al.
Cellular microbiology, 16(7), 1080-1093 (2014-02-14)
Members of the interferon-induced transmembrane (IFITM) protein family inhibit the entry of a wide range of viruses. Viruses often exploit the endocytosis pathways to invade host cells and escape from the endocytic vesicles often in response to low pH. Localization
Validation of a method for preparing influenza H5N1 simulated samples.
John A Lednicky,Julie M Villanueva,Stephen A Burke,Roxanne Shively,Michael W Shaw et al.
Journal of Virological Methods null
Shuguang Han et al.
Clinical and experimental pharmacology & physiology, 45(1), 84-93 (2017-08-31)
Curcumin, an active phenolic agent extract from the Curcuma longa, exhibits excellent anti-cancer, anti-inflammation, and neuroprotective effects. We aimed to investigate the anti-influenza role of curcumin in vitro and in vivo. The effect of curcumin on replication of influenza A
Donald D Raymond et al.
Nature medicine, 22(12), 1465-1469 (2016-11-08)
For broad protection against infection by viruses such as influenza or HIV, vaccines should elicit antibodies that bind conserved viral epitopes, such as the receptor-binding site (RBS). RBS-directed antibodies have been described for both HIV and influenza virus, and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service