Skip to Content
Merck
All Photos(1)

Documents

MAB5550

Sigma-Aldrich

Anti-TorsinA Antibody

ascites fluid, Chemicon®

Synonym(s):

Anti-AMC5, Anti-DYT1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

monoclonal

species reactivity

mouse, rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TOR1B(27348)

General description

TorsinA can regulate the cellular trafficking of the dopamine transporter, as well as other polytopic membrane-bound proteins, including G protein-coupled receptors, transporters, and ion channels. TorsinA is found in the dopamine neurons that are lost in PD, and is a component of Lewy bodies. in addition, a genetic defect in TorsinA underlies Torsion dystonia, a disease characterized by painful muscle contractions that result in uncontrollable distortions.

Specificity

TorsinA

Immunogen

Maltose binding protein fusion protein from human torsinA.

Application

Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Neurodegenerative Diseases
This Anti-TorsinA Antibody is validated for use in IP, WB, IC for the detection of TorsinA.

Target description

37kDa

Physical form

Ascites
Liquid. Contaings no preservative

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months after date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Positive Control: Human brain lysate, NGF treated PC12 cells.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diminishing evidence for torsinA-positive neuronal inclusions in DYT1 dystonia.
Drew Pratt et al.
Acta neuropathologica communications, 4(1), 85-85 (2016-08-18)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service