Skip to Content
Merck
All Photos(3)

Key Documents

MAB5224

Sigma-Aldrich

Anti-Internexin α Antibody, CT, clone 2E3

culture supernatant, clone 2E3, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

culture supernatant

antibody product type

primary antibodies

clone

2E3, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... INA(9118)

General description

The protein alpha-internexin is a ~66 kDa Class IV intermediate filament originally discovered as it copurifies with other neurofilament subunits {Pachter J et al J. Cell Biol 101:1316-22 (1985). It is related to but distinct from the better known neurofilament triplet proteins, NF-L, NF-M and NF-H, having similar protein sequence motifs and a similar intron organization. It is expressed in large amounts early in neuronal development, but is downregulated in many neurons as development procedes. Many classes of mature neurons contain alpha-internexin in addition to NF-L, NF-M anbd NF-H. In some mature neurons alpha-internexin is the only neurofilament subunit expressed. Antibodies to a-internexin are therefore unique probes to study and classify neuronal types and follow their processes in sections and in tissue culture. In addition the very early developmental expression of alpha-internexin means its presence is an early and convenient diagnostic feature of neuronal progenitors cells and other cell committed to the neuronal lineage. In addition recent studies show a marked up-regulation of a-internexin during neuronal regeneration {McGraw et al 2002}. The use of antibodies to this protein in the study of brain tumors has not been examined to date, but is likely to be of interest.

Specificity

Alpha internexin. Localized to the C-terminal 164 amino acids of rat alpha internexin.

Immunogen

Epitope: C-terminus
Full length recombinant rat alpha internexin.

Application

Research Category
Neuroscience
Research Sub Category
Neurofilament & Neuron Metabolism

Neuronal & Glial Markers
This Anti-Internexin Antibody, α, C-terminus, clone 2E3 is validated for use in ELISA, IP, WB, IH(P) for the detection of Internexin.
Western blot: antibody detects a protein band at 55-66kDa, as expected for alpha internexin {Evans, et al, 2001}.

Immunocytochemistry on PC12 and Ntera-2 cells.

Immunohistochemistry on formalin and methanol fixed tissues. Works on paraffin embedded tissue sections.

Immunoprecipitation

ELISA

Optimal working dilutions must be determined by the end user.

Linkage

Replaces: 04-1032

Physical form

Mouse IgG in low serum (1%), tissue culture supernatant containing no preservative.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mala V Rao et al.
Journal of neurochemistry, 137(2), 253-265 (2016-01-13)
Amyotrophic lateral sclerosis (ALS) is a progressive motor neuron disease with a poorly understood cause and no effective treatment. Given that calpains mediate neurodegeneration in other pathological states and are abnormally activated in ALS, we investigated the possible ameliorative effects
Aidong Yuan et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(36), 11316-11329 (2009-09-11)
The ultrastructural view of the axonal cytoskeleton as an extensively cross-linked network of neurofilaments (NFs) and other cytoskeletal polymers contrasts with the dynamic view suggested by axonal transport studies on cytoskeletal elements. Here we reconcile these perspectives by showing that
Deepti Narasimhaiah et al.
Neuropathology : official journal of the Japanese Society of Neuropathology, 32(1), 30-37 (2011-04-13)
Recently, mutations in IDH1 and IDH2 have been reported as an early and common genetic alteration in diffuse gliomas, being possibly followed by 1p/19q loss in oligodendrogliomas and TP53 mutations in astrocytomas. Lately, IDH1 mutations have also been identified in
Wei-Hao Peng et al.
Frontiers in neuroscience, 15, 728905-728905 (2021-11-23)
Purpose: The rearranged during transfection (RET) receptor tyrosine kinase plays a key role in transducing signals related to cell growth and differentiation. Ret mutant mice show abnormal retinal activity and abnormal levels and morphology of bipolar cells, yet die on
F N Ko et al.
The Journal of clinical investigation, 98(6), 1493-1501 (1996-09-15)
The mitogenic effect of activated coagulation factor X (factor Xa) was examined in cultured aortic smooth muscle cells (VSMC) from Wistar-Kyoto rats (WKY). Factor Xa stimulated DNA synthesis and cell growth in VSMC, not through the phospholipase C-protein kinase C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service