Skip to Content
Merck
All Photos(1)

Documents

MAB4383

Sigma-Aldrich

Anti-Nuclei Antibody, clone 3E1.3

clone 3E1.3, Chemicon®, from mouse

Synonym(s):

Anti-Nuclei, Clone 3E1.3 Anti-Nuclei, Nuclei Detection Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3E1.3, monoclonal

species reactivity

human

should not react with

mouse

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable

input

sample type neural stem cell(s)

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

General description

Stains nuclei of all human cell types giving a diffuse nuclear staining pattern. Chromosomes are negatively stained in metaphase cells.

Immunogen

Purified Recombinant Human Nuclei

Application

Anti-Nuclei Antibody, clone 3E1.3 detects level of Nuclei & has been published & validated for use in FC, IC, IH.
Research Category
Stem Cell Research
Research Sub Category
Neural Stem Cells

Physical form

Format: Purified
Liquid in PBS with 0/1% sodium azide

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weizhe Li et al.
Disease models & mechanisms, 10(12), 1489-1502 (2017-11-01)
Studies of innate glial cell responses for progressive human immunodeficiency virus type one (HIV-1) infection are limited by a dearth of human disease-relevant small-animal models. To overcome this obstacle, newborn NOD/SCID/IL2Rγc-/- (NSG) mice were reconstituted with a humanized brain and
Jyoti Kode et al.
The Indian journal of medical research, 139(4), 585-597 (2014-06-14)
In current era of limb-salvage therapy, pasteurization of bone sarcomas is receiving growing attention as a potential extracorporeal treatment and cost-effective alternative to allografts and radiation before surgical reimplantation. Detailed in vitro and in vivo pre-clinical study to evaluate efficacy
Ruslan L Nuryyev et al.
Cell transplantation, 26(11), 1811-1821 (2018-01-18)
The use of regenerative medicine to treat nervous system disorders like ataxia has been proposed to either replace or support degenerating neurons. In this study, we assessed the ability of human neural progenitor cells (hNPCs) to repair and restore the
Qian Li et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(9), 2375-2387 (2014-05-03)
Glioblastoma is the most common adult primary malignant intracranial cancer. It is associated with poor outcomes because of its invasiveness and resistance to multimodal therapies. Human adipose-derived mesenchymal stem cells (hAMSC) are a potential treatment because of their tumor tropism
Virginie Lamour et al.
International journal of cancer, 137(5), 1047-1057 (2015-01-27)
Osteopontin (OPN) is a secreted protein involved in most aspects of tumor progression and metastasis development. Elevated OPN expression has been reported in multiple types of cancer including glioblastoma (GBM), the highest grade and most aggressive brain tumor. GBMs contain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service