Skip to Content
Merck
All Photos(1)

Documents

MAB4310

Sigma-Aldrich

Anti-CD133 Antibody, clone 13A4

clone 13A4, Chemicon®, from rat

Synonym(s):

Prominin-1, AC133 (human)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

13A4, monoclonal

species reactivity

mouse

should not react with

chicken, Drosophila, rat

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

input

sample type hematopoietic stem cell(s)
sample type neural stem cell(s)
sample type epithelial cells
sample type: mouse embryonic stem cell(s)
sample type mesenchymal stem cell(s)

isotype

IgG1κ

suitability

not suitable for immunohistochemistry (Paraffin)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

dog ... Prom1(488816)
human ... PROM1(8842)
mouse ... Prom1(19126)
rat ... Prom1(60357)

Related Categories

General description

Prominin is a novel plasma membrane protein with an N-terminal extracellular domain, five transmembrane segments flanking two short cytoplasmic loops and two large glycosylated extracellular domains, and a cytoplasmic C-terminal domain. DNA sequences from Caenorhabditis elegans predict the existence of a protein with the same features, suggesting that prominin is conserved between vertebrates and invertebrates. Prominin is found not only in the neuroepithelium but also in various other epithelia of the mouse embryo. In the adult mouse, prominin has been detected in the brain ependymal layer, and in kidney tubules. In these epithelia, prominin is specific to the apical surface, where it is selectively associated with microvilli and microvilli-related structures. Remarkably, upon expression in CHO cells, prominin is preferentially localized to plasma membrane protrusions such as filopodia, lamellipodia, and microspikes. These observations imply that prominin contains information to be targeted to, and/or retained in, plasma membrane protrusions rather than the planar cell surface. Moreover, results show that the mechanisms underlying targeting of membrane proteins to microvilli of epithelial cells and to plasma membrane protrusions of non-epithelial cells are highly related. {Weigmann, A et al.1997}. Also see {http://www.ncbi.nlm.nih.gov/prow/guide/438375806_g.htm} for more information on CD133 and prominin expression.

Specificity

Recognizes mouse Prominin-1 (sometimes also referred to as CD133 and, in the case of the human orthologue, as AC133), a 115-120 kDa pentaspan transmembrane (5-TM) domain glycoprotein. Prominin-1 is expressed on primitive cells such as hematopoietic stem and progenitor cells, neural & endothelial stem cells, retina and retinoblastoma, as well as developing epithelium. To date, the function and ligand of Prominin-1 are unknown.

This antibody can be used to isolate murine stem cells from brain and bone marrow.

The 13A4 antibody does not cross react with rat, human, chicken, or Drosophila antigen. The original paper is located at: http://www.pnas.org/cgi/reprint/94/23/12425.pdf

Immunogen

Telencephali of ten 12-day-old NMRI mouse embryos (E12, ~1 mg protein), which consist mostly of neuroepithelium, were homogenized in 1 ml PBS, mixed with crushed nitrocellulose filter (1 cm2 in 0.5 ml PBS) as adjuvant, and injected i.p. into a LouXSD rat. {Weigmann et al, PNAS 1997}

Application

Anti-CD133 Antibody, clone 13A4 detects level of CD133 & has been published & validated for use in FC, IH, IP & WB.

Western Blot Analysis: A 1:1000 dilution from a representative lot detected CD133 in monkey kidney lysate.
Research Category
Stem Cell Research
Research Sub Category
Neural Stem Cells

Hematopoietic Stem Cells

Target description

115 kDa

Physical form

Format: Purified
Protein A Purified mouse immunoglobulin in 20 mM sodium phosphate, 250 mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative.
Protein A purified

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Positive Control: Kidney tissue or membranes
Negative Control: Liver tissue or membranes

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taofeek K Owonikoko et al.
Journal of translational medicine, 14(1), 111-111 (2016-05-05)
SCLC has limited treatment options and inadequate preclinical models. Promising activity of arsenic trioxide (ASO) recorded in conventional preclinical models of SCLC supported the clinical evaluation of ASO in patients. We assessed the efficacy of ASO in relapsed SCLC patients
Regional differences in stem cell/progenitor cell populations from the mouse achilles tendon.
Mienaltowski, MJ; Adams, SM; Birk, DE
Tissue Engineering: Part A null
Zhibo Xia et al.
Molecular cancer research : MCR, 11(9), 1101-1111 (2013-06-14)
Glioma-initiating cells (GIC), which are characterized by their self-renewal capacity and tumorigenicity, were recently identified as a highly tumorigenic subpopulation of glioblastoma multiforme and are considered responsible for glioblastoma recurrence and chemo/radiation resistance. Previously, it was revealed that Wnt signaling
Activation of the sonic hedgehog signaling pathway occurs in the CD133 positive cells of mouse liver cancer Hepa 1-6 cells.
Jeng, KS; Sheen, IS; Jeng, WJ; Yu, MC; Hsiau, HI; Chang, FY; Tsai, HH
OncoTargets and Therapy null
Ectopic expression of the WWOX gene suppresses stemness of human ovarian cancer stem cells.
Yan, HC; Xu, J; Fang, LS; Qiu, YY; Lin, XM; Huang, HX; Han, QY
Oncology Letters null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service