Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
mouse
Quality Segment
conjugate
unconjugated
antibody form
ascites fluid
clone
ST126, monoclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
neutralization: suitable
isotype
IgG2b
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Immunogen
Application
Dilute with buffer pH 7.5 - 8.0 to desired working volumes. Final working dilutions must be determined by end user. For extensive dilution use protein-containing or other stabilizing medium.
Inflammation & Immunology
Cytokines & Cytokine Receptors
Biochem/physiol Actions
Physical form
Preparation Note
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description ascites fluid, clone ST126, Chemicon® | description clone 37895, purified immunoglobulin, lyophilized powder | description clone MMHAR-2, Chemicon®, from mouse | description serum, Chemicon® |
| species reactivity human | species reactivity mouse | species reactivity human | species reactivity human |
| biological source mouse | biological source rat | biological source mouse | biological source rabbit |
| clone ST126, monoclonal | clone 37895, monoclonal | clone MMHAR-2, monoclonal | clone polyclonal |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form ascites fluid | antibody form purified immunoglobulin | antibody form purified immunoglobulin | antibody form serum |
| technique(s) neutralization: suitable | technique(s) flow cytometry: suitable, western blot: suitable, neutralization: suitable | technique(s) immunoprecipitation (IP): suitable, flow cytometry: suitable, immunohistochemistry: suitable | technique(s) immunohistochemistry: suitable, neutralization: suitable, ELISA: suitable, immunoprecipitation (IP): suitable, western blot: suitable |
Storage Class
10 - Combustible liquids
WGK
WGK 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



