Skip to Content
Merck
All Photos(1)

Key Documents

MAB411

Sigma-Aldrich

Anti-Interferon-α2 Antibody, clone ST2541

ascites fluid, clone ST2541, Chemicon®

Synonym(s):

Interferon alpha-2, IFN-alpha-2, Interferon alpha-A, LeIF A

Sign Into View Organizational & Contract Pricing

Select a Size

500 μL
€370.00

€370.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
500 μL
€370.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€370.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

ST2541, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
affinity binding assay: suitable
neutralization: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... IFNA2(3440)

General description

Interferon alpha-2 (UniProt P01563; also known as IFN-alpha-2, Interferon alpha-A, LeIF A) is encoded by the IFNA2 (also known as IFNA2A, IFNA2B, IFNA2C) gene (Gene ID 3440) in human. Interferons (IFNs) are cytokines whose prouction and release by host cells are induced in the presence of pathogens (e.g. viruses, bacteria, parasites) and tumor cells. Originally named for their ability to interfere with viral replication, IFNs mediate the activate immune cells, such as natural killer cells and macrophages, and increase host defense by up-regulating antigen presentation by boosting the expression of major histocompatibility complex (MHC) antigens. More than twenty distinct IFN genes and proteins have been identified and classified into three main types. Type I IFNs (IFN-α, IFN-β, IFN-ε, IFN-κ, and IFN-ω) exert their function via binding IFN-α/β receptor (encoded by the IFNAR1 and IFNAR2 gene, respectively). IFN-γ is the only known type II IFN, it is released by type 1 T helper cells and binds IFN-γ receptor (encoded by the IFNGR1 and IFNGR2 gene, respectively). Type III IFNs signal through a receptor complex consisting of IL10R2 (CRF2-4) and IFNLR1 (CRF2-12).

Specificity

Clone ST2541 neutralized human IFN-αA (IFN-α2) and IFN-αN (produced by Nalmalwa human Burkitt′s lymphoma cells), but not IFN-α1, antiviral activity (Shearer, M., et al. (1984). J. Immunol. 133(6):3096-3101).

Immunogen

Human alpha-interferon.

Application

Detect Interferon-α2 using this Anti-Interferon-α2 Antibody, clone ST254 validated for use in NEUT, WB, ELISA, Affinity binding.
Neutralizing Analysis: A representative lot enhanced viral replication by neutralizing host-secreted IFN in human prostate cancer PC3 cultures infected with P/V-CPI(−) and P/V-CPI(−)-G3A at a low MOI (Gainey, M.D., et al. (2000). J. Virol. 82(19):9369-9380).
Neutralizing Analysis: A representative lot neutralized human IFN-αA (IFN-α2) and IFN-αN (produced by Nalmalwa human Burkitt′s lymphoma cells), but not IFN-α1, antiviral activity against Semliki Forest Virus in human WISH amniotic epithelial cell cultures (Shearer, M., et al. (1984). J. Immunol. 133(6):3096-3101).
Affinity Binding Assay: A representative lot captured human IFN-α2, but not human IFN-α1 (Shearer, M., et al. (1984). J. Immunol. 133(6):3096-3101).
ELISA Neutralizing Analysis: A representative lot was used as the detection antibody for the detection of human IFN-αN (produced by Nalmalwa human Burkitt′s lymphoma cells) by sandwich ELISA (Shearer, M., et al. (1984). J. Immunol. 133(6):3096-3101).
Western Blotting Analysis: A representative lot detected IFN-αA (IFN-α2) and IFN-αN (produced by Nalmalwa human Burkitt′s lymphoma cells) under reducing condition by Western blotting (Shearer, M., et al. (1984). J. Immunol. 133(6):3096-3101).
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors

Physical form

Liquid, no preservatives

Storage and Stability

Store at -20°C for up to one year. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Innate immune signaling induces high levels of TC-specific deaminase activity in primary monocyte-derived cells through expression of APOBEC3A isoforms.
Thielen, BK; McNevin, JP; McElrath, MJ; Hunt, BV; Klein, KC; Lingappa, JR
The Journal of Biological Chemistry null
Francesca Marino-Merlo et al.
Cell death & disease, 7(9), e2354-e2354 (2016-09-02)
The transcription factor nuclear factor-kappa B (NF-κB) is a crucial player of the antiviral innate response. Intriguingly, however, NF-κB activation is assumed to favour herpes simplex virus (HSV) infection rather than restrict it. Apoptosis, a form of innate response to
Jonathan C Reed et al.
The Journal of cell biology, 198(3), 439-456 (2012-08-02)
To produce progeny virus, human immunodeficiency virus type I (HIV-1) Gag assembles into capsids that package the viral genome and bud from the infected cell. During assembly of immature capsids, Gag traffics through a pathway of assembly intermediates (AIs) that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service