Skip to Content
Merck
All Photos(1)

Key Documents

MAB397A4

Sigma-Aldrich

Anti-GluR2 Antibody, clone 6C4, Alexa Fluor 488 Conjugate

clone 6C4, from mouse, ALEXA FLUOR 488

Synonym(s):

Glutamate receptor 2, GluR-2, AMPA-selective glutamate receptor 2, GluR-B, GluR-K2, Glutamate receptor ionotropic, AMPA 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€557.00

€557.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€557.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€557.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6C4, monoclonal

species reactivity

mouse, rat

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GRIA2(2891)

General description

Glutamate receptors (GluRs) can be categorized as ionotropic or metabotropic and subcatergorized by their agonist preferences (NMDA, AMPA or Kainic acid). There are four types of AMPA selective GluR subunits (GluR1, GluR2, GluR3 and GluR4). Tetrameric or pentameric combinations of different subunits contributes to the functional diversity of AMPA receptors. In general, AMPA receptors mediate fast synaptic current at most excitatory synapses, with stoichiometry characterized by subtype composition. Although subunit composition of AMPA receptors varies, they must contain at least one edited GluR2 subunit to be calcium impermeable. The critical residue controlling calcium permeability is in the pore loop region. In GluR1, GluR3, and GluR4, this positionis occupied by a Gln residue. In GluR2, it is occupied by an Arg residue. It has been shown experimentally that the presence of Arg in this position blocks CA2+ ion permeability, while a Gln does not. Relative calcium permeability in AMPA receptor channels may be significant in pathological neurotoxic damage and long term changes in nervous system responses.

Specificity

Recognizes the large N-terminal extracellular domain of Glutamate Receptor 2 (GluR2). No cross-reactivity observed with other AMPA/Kainate GluR subunits.

Immunogen

Epitope: Extracellular domain
Recombinant fusion protein TrpE-GluR2

Application

Immunocytochemistry analysis: A 1:100 dilution from a representative lot detected GluR2 in rat E18 primary cortex cells.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
This Anti-GluR2 Antibody, clone 6C4, Alexa Fluor 488 Conjugate is validated for use in IH, IC for the detection of GluR2.

Quality

Evaluated by Immunohistochemistry in adult mouse brain tissue.

Immunohistochemistry Analysis: A 1:100 dilution of this antibody detected GluR2 in adult mouse brain tissue.

Target description

The unconjugated parent antibody (Cat. No. MAB397) has an observed molecular weight at ~102 kDa

Physical form

Protein A purified
Purified mouse monoclonal IgG2a conjugated to Alexa Fluor 488 in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Adult mouse brain tissue

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boris Brant et al.
Molecular psychiatry (2021-09-19)
Mutations in the IQSEC2 gene are associated with drug-resistant, multifocal infantile and childhood epilepsy; autism; and severe intellectual disability (ID). We used induced pluripotent stem cell (iPSC) technology to obtain hippocampal neurons to investigate the neuropathology of IQSEC2-mediated disease. The
Eli J Rogers et al.
Frontiers in molecular neuroscience, 12, 43-43 (2019-03-08)
We have recently described an A350V mutation in IQSEC2 associated with intellectual disability, autism and epilepsy. We sought to understand the molecular pathophysiology of this mutation with the goal of developing targets for drug intervention. We demonstrate here that the
G Aleph Prieto et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 37(5), 1197-1212 (2016-12-18)
Long-term potentiation (LTP) is an activity-dependent and persistent increase in synaptic transmission. Currently available techniques to measure LTP are time-intensive and require highly specialized expertise and equipment, and thus are not well suited for screening of multiple candidate treatments, even

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service