Skip to Content
Merck
All Photos(1)

Key Documents

MAB390

Sigma-Aldrich

Anti-p75 LNGFR Antibody, Saporin conjugated, clone 192

clone 192, Chemicon®, from mouse

Synonym(s):

CD271 antigen, Low affinity neurotrophin receptor p75NTR, Low-affinity nerve growth factor receptor, NGF receptor, low affinity nerve growth factor receptor, nerve growth factor receptor, nerve growth factor receptor (TNFR superfamily, member 16), p75 IC

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€152.00

€152.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€152.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€152.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

saporin

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

192, monoclonal

species reactivity

rat

should not react with

human, mouse

packaging

antibody small pack of 25 μg

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NGFR(4804)

General description

The Low Affinity Nerve Growth Factor Receptor (also called the LNGFR or p75 neurotrophin receptor) is one of the two receptor types for the neurotrophins, a family of protein growth factors which stimulate neuronal cells to survive and differentiate. The neurotrophins are comprised of four proteins, all of which bind to the LNGFR: Nerve Growth Factor (NGF), Brain Derived Neurotrophic Factor (BDNF), Neurotrophin-3 (NT-3), and Neurotrophin-4 (NT-4. Nerve growth factor, the prototypical growth factor, is a protein secreted by a neuron′s target. NGF is critical for the survival and maintenance of sympathetic and sensory neurons. NGF is released from the target cells, binds to and activates its high affinity receptor (TrkA), and is internalized into the responsive neuron. The NGF/TrkA complex is subsequently trafficked back to the cell body. This movement of NGF from axon tip to soma is thought to be involved in the long-distance signaling of neurons.

Specificity

Recognizes the rat NGF receptor (p75, low affinity Neurotrophin Receptor). MAB365 is the monoclonal antibody used for MAB390-SAP.

Immunogen

p75-expressing cells

Application

Immunohistochemistry (Paraffin):
A previous lot of this antibody was used in IH(P).

Immunocytochemistry:
A previous lot of this antibody was used in IC.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Neurochemistry & Neurotrophins
This Anti-p75 Low Affinity Nerve Growth Factor Receptor Antibody, Saporin conjugated, clone 192 is validated for use in ICC, IHC(P), FUNC for the detection of p75 Low Affinity Nerve Growth Factor Receptor.

Physical form

Protein A purified
Saporin Conjugated. 192-IgG-SAP is sterile filtered and presented in Dulbecco′s PBS with no preservative.

Storage and Stability

Centrifuge material at low speed in microfuge to ensure all of the solution is at the bottom of the tube. Vortex gently. Long term storage at -70ºC or below. Short term storage at -20ºC or 2-8ºC with appropriate antibacterial agent if compatible with the intended application. The sterile conjugate is stable at 2-8ºC for several months. Avoid repeated freezing and thawing. Glycerol (1:1) can be added for additional stability if compatible with the intended application. This immunotoxin may be handled safely using standard laboratory procedures as the cross-species reactivity between these anti-rat p75 antibodies and the human form of p75 is negligible.

Analysis Note

Control
Brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christiana M Cooper-Kuhn et al.
Journal of neuroscience research, 77(2), 155-165 (2004-06-24)
Adult neurogenesis has been shown to be regulated by a multitude of extracellular cues, including hormones, growth factors, and neurotransmitters. The cholinergic system of the basal forebrain is one of the key transmitter systems for learning and memory. Because adult
James M Conner et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(35), 10883-10889 (2009-09-04)
Nerve growth factor (NGF) is produced in the hippocampus throughout life and is retrogradely trafficked to septal cholinergic neurons, providing a potential mechanism for modulating cholinergic inputs and, thereby, hippocampal plasticity. To explore NGF modulation of hippocampal plasticity and function
Carlos Blanco-Centurion et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 27(51), 14041-14048 (2007-12-21)
The hypocretin (HCRT) neurons are located only in the perifornical area of the lateral hypothalamus and heavily innervate the cholinergic neurons in the basal forebrain (BF), histamine neurons in the tuberomammillary nucleus (TMN), and the noradrenergic locus ceruleus (LC) neurons
Nicole C Berchtold et al.
Journal of neuroscience research, 68(5), 511-521 (2002-07-12)
Brain-derived neurotrophic factor (BDNF) enhances synaptic plasticity and neuron function. We have reported that voluntary exercise increases BDNF mRNA levels in the hippocampus; however, mechanisms underlying this regulation have not been defined. We hypothesized that medial septal cholinergic and/or gamma
Else A Tolner et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(2), 692-702 (2012-01-13)
Patterned neuronal activity such as spindle bursts in the neonatal cortex is likely to promote the maturation of cortical synapses and neuronal circuits. Previous work on cats has shown that removal of subplate neurons, a transient neuronal population in the

Questions

  1. Why are there multiple bands on a western blot for the p75 antbody?

    1 answer
    1. This antibody is validated only for immunohistochemistry and immunocytochemistry. Western blotting is not a recommended technique for this antibody, making it difficult to predict the nature of the bands in Western blotting results.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service