Skip to Content
Merck
All Photos(2)

Key Documents

MAB3839-I

Sigma-Aldrich

Anti-Transglutaminase-2 Antibody, FN binding domain Antibody, clone 4G3

clone 4G3, from mouse

Synonym(s):

Protein-glutamine gamma-glutamyltransferase 2, EC: 2.3.2.13, Tissue transglutaminase, Transglutaminase C, TG(C), TGC, TGase C, Transglutaminase H, TGase H, TGase-2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€154.00
100 μG
€403.00

€154.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€154.00
100 μG
€403.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€154.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4G3, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... TGM2(7052)

General description

Protein-glutamine gamma-glutamyltransferase 2 (UniProt: P21980; also known as EC: 2.3.2.13, Tissue transglutaminase, Transglutaminase C, TG(C), TGC, TGase C, Transglutaminase H, TGase H, Transglutaminase-2, TGase-2, TG2) is encoded by D13the TGM2 gene (Gene ID: 7052) in human. TG2 is a multi-domain, multi-functional enzyme that is shown to post-translationally modify proteins by catalyzing the formation of intermolecular isopeptide bonds between glutamine and lysine side-chains. It catalyes the calcium-dependent covalent modification of protein-bound glutamine side chains through transamidation or deamidation reactions. Recycling endosomes is reported to be essential for externalization of TG2, which does not follow the classical ER/Golgi-dependent secretion pathway, but is targeted to perinuclear recycling endosomes, and is delivered inside these vesicles prior to externalization. Inactivation of recycling endosomes or blocking endosome fusion with the plasma membrane is shown to block TG2 secretion. TG2 plays a critical role in extracellular matrix formation and integrin mediated signaling. It is also reported to be involved in cell signaling involving 7-transmembrane receptors. It also participates in the pathogenesis of neurodegenerative diseases and certain cancers. (Ref.: Zemskov, EA., et al (2011). PLoS ONE 6(4): e19414; Siegal, M., and Khosla, C. (2007). Pharmacol. Ther. 115(2); 232-245.

Specificity

Clone 4G3 specifically detects human Transglutaminase-2.

Immunogen

A full-length purified tissue transglutaminase (TG-2) from human erythrocytes.

Application

Anti-Transglutaminase-2, FN binding domain, clone 4G3, Cat. No. MAB3839-I, is a mouse monoclonal antibody that detects Protein-glutamine gamma-glutamyltransferase 2 and has been tested for use in Flow Cytometry, Immunofluorescence, Immunoprecipitation, and Western Blotting.
Research Category
Signaling
Western Blotting Analysis: 4 µg/mL from a representative lot detected Transglutaminase-2, FN binding domain in HUVEC lysate.

Western Blotting Analysis: A representative lot detected Transglutaminase-2, FN binding domain in Western Blotting applications (Janiak, A., et. al. (2006). Mol Biol Cell. 17(4):1606-19).

Radioimmunoassay Analysis: A representative lot detected Transglutaminase-2, FN binding domain in Radioimmunoassay applications (Akimov, S.S., et. al. (2001). Blood. 98(5):1567-76).

Flow Cytometry Analysis: A representative lot detected Transglutaminase-2, FN binding domain in Flow Cytometry applications (Zemskov, E.A., et. al. (2011). PLos One. 6(4):e19414).

Immunoprecipitation Analysis: A representative lot immunoprecipitated Transglutaminase-2, FN binding domain in Immunoprecipitation applications (Zemskov, E.A., et. al. (2011). PLos One. 6(4):e19414; Janiak, A., et. al. (2006). Mol Biol Cell. 17(4):1606-19).

Immunofluorescence Analysis: A representative lot detected Transglutaminase-2, FN binding domain in Immunofluorescence applications (Janiak, A., et. al. (2006). Mol Biol Cell. 17(4):1606-19; Akimov, S.S., et. al. (2001). Blood. 98(5):1567-76; Zemskov, E.A., et. al. (2011). PLos One. 6(4):e19414).

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Transglutaminase-2, FN binding domain in A431 cell lysate.

Target description

~80 kDa observed; 77.33 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: MAB3839

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service