Skip to Content
Merck
All Photos(5)

Documents

MAB3757-I

Sigma-Aldrich

Anti-VEGF Receptor-3 Antibody, clone 9D9F9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.74

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

9D9F9, monoclonal

mol wt

calculated mol wt 152.76 kDa
observed mol wt ~190 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μL

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

human ... FLT4(2324)

General description

Vascular endothelial growth factor receptor 3 (UniProt: P35916; also known as EC:2.7.10.1, VEGFR-3, Fms-like tyrosine kinase 4, FLT-4, Tyrosine-protein kinase receptor FLT4) is encoded by the FLT4 (also known as VEGFR3) gene (Gene ID: 2324) in human. FLT-4 is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-24), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 25-775), a transmembrane domain (aa 776-796), and cytoplasmic domain (aa 797-1363). It is expressed in endothelial cells and is also detected in fetal spleen, lung, and brain. In adults it is detected in liver, muscle, thymus, placenta, lung, testis, ovary, prostate, heart, and kidney. FLT-4 has tyrosine-protein kinase activity and acts as a receptor for VEGFC and VEGFD. It plays a role in adult lymphangiogenesis and in the development of the vascular network and the cardiovascular system during embryonic development. Its secreted form functions as a decoy receptor for VEGFC and/or VEGFD and play an important role as a negative regulator of VEGFC-mediated lymphangiogenesis and angiogenesis. Its major function is to promote proliferation, survival, and migration of endothelial cells, and regulates angiogenic sprouting. FLT-4 undergoes trans auto-phosphorylation on tyrosine residues upon ligand binding. Phosphorylation at tyrosine 1068 is required for autophosphorylation at additional tyrosine residues. Phosphorylation at tyrosine 1063 and 1337 are important for interaction with Crk adaptor proteins and subsequent activation of MAPK8. In addition, its phosphorylation at tyrosine 1230, 1231, and 1337 are reported to be important for interaction with Grb2 and subsequent activation of the AKT1 and ERK1/ERK2 signaling pathways.

Specificity

Clone 9D9F9 is a mouse monoclonal antibody that detects Vascular endothelial growth factor receptor 3 (VEGFR-3). It targets an epitope within the extracellular domain.

Immunogen

A recombinant fragment corresponding to the extracellular domain of human Vascular endothelial growth factor receptor 3 (VEGFR-3).

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from HEK293 cells over-expressing VEGFR-3.

Western Blotting Analysis (WB): A 1:500 dilution of this antibody detected Vascular endothelial growth factor receptor 3 (VEGFR-3) in lysate from HEK293 cells over-expressing human VEGFR-3.

Tested Applications

Immunoprecipitation Analysis: A representative lot immunoprecipitated VEGF Receptor-3 in Immunoprecipitation applications (Bando, H., et al. (2004). Int J Cancer; 111(2):184-91).

ELISA Analysis: A representative lot detected VEGF Receptor-3 in ELISA applications (Bando, H., et al. (2004). Int J Cancer; 111(2):184-91).

Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected VEGF Receptor-3 in human placenta and human kidney tissue sections.

Western Blotting Analysis: A representative lot detected VEGF Receptor-3 in Western Blotting applications (Bando, H., et al. (2004). Int J Cancer; 111(2):184-91; Zhang, Y., et al. (2018). Nat Commun.; 9(1):1296).

Immunohistochemistry Applications: A representative lot detected VEGF Receptor-3 in Immunohistochemistry applications (Bando, H., et al. (2004). Int J Cancer; 111(2):184-91; Jussila, L. et al. (1998). Cancer Res.; 58(8):1599-604; Su, C., et al. (2011). J Exp Clin Cancer Res.; 30(1):85; Valtola, R., et al. (1999). Am J. Pathol.; 154(5):1381-90).

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected VEGF Receptor-3 in human umbilical vascular endothelial cells (HUVEC).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-VEGF Receptor-3, clone 9D9F9, Cat. No. MAB3757-I, is a mouse monoclonal antibody that detects VEGF receptor 3 and is tested for use in ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service