Skip to Content
Merck
All Photos(1)

Key Documents

MAB338

Sigma-Aldrich

Anti-Ciliary Neurotrophic Factor Antibody, clone 4-68

clone 4-68, Chemicon®, from mouse

Synonym(s):

CNTF

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€753.00

€753.00


Estimated to ship on06 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€753.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€753.00


Estimated to ship on06 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4-68, monoclonal

species reactivity

mouse, rat, human, monkey

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CNTF(1270)

Specificity

Ciliary neurotrophic factor. Detects a 22 kD band in Western blots of rat sciatic nerve extract.

Immunogen

Purified ciliary neurotrophic factor from rat sciatic nerve.

Application

Immunohistochemistry: 10-20 μg/mL on frozen sections of rat sciatic nerve perfused and counter-fixed with 4% paraformaldehyde (Stockli et al., 1991).

Western blotting: 10 μg/mL (Stockli et al., 1991)

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins
This Anti-Ciliary Neurotrophic Factor Antibody, clone 4-68 is validated for use in WB, IH for the detection of Ciliary Neurotrophic Factor.

Physical form

Format: Purified
Liquid in 0.02M phosphate buffer, 0.25M NaCl with 0.1% sodium azide, pH 7.6.

Storage and Stability

Maintain at 2-8°C for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cuihong Jia et al.
Glia, 66(11), 2456-2469 (2018-12-01)
Astrocyte-derived ciliary neurotrophic factor (CNTF) promotes adult subventricular zone (SVZ) neurogenesis. We found that focal adhesion kinase (FAK) and JNK, but not ERK or P38, repress CNTF in vitro. Here, we defined the FAK-JNK pathway and its regulation of CNTF
Milena Girotti et al.
Neuropharmacology, 160, 107791-107791 (2019-09-26)
Deficits in cognitive flexibility, i.e. the ability to modify behavior in response to changes in the environment, are present in several psychiatric disorders and are often refractory to treatment. However, improving treatment response has been hindered by a lack of
Anna Polosa et al.
International journal of molecular sciences, 20(11) (2019-06-07)
To unravel the mechanisms behind the higher resistance to light damage of juvenile (JR) versus adult (AR) rats, Sprague Dawley rats were exposed to a bright luminous environment of 10, 000 lux. The light-induced retinopathy (LIR) was assessed with histology
Cuihong Jia et al.
Stem cell research, 49, 102061-102061 (2020-11-02)
Constant neuroregeneration in adult olfactory epithelium maintains olfactory function by basal stem cell proliferation and differentiation to replace lost olfactory sensory neurons (OSNs). Understanding the mechanisms regulating this process could reveal potential therapeutic targets for stimulating adult olfactory neurogenesis under
Laurène Cagnon et al.
Neurobiology of disease, 33(1), 133-142 (2008-11-11)
In pediatric patients, hyperammonemia can provoke irreversible damages to developing CNS like cortical atrophy, ventricular enlargement, demyelination or gray and white matter hypodensities which are concordant with alterations of neurons and oligodendrocytes. Cerebral injury triggers endogenous protective mechanisms that can

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service