Skip to Content
Merck
All Photos(1)

Key Documents

MAB2510

Sigma-Aldrich

Anti-Green Fluorescent Protein Antibody, original GFP only, clone 264-449-2

clone 264-449-2, Chemicon®, from mouse

Synonym(s):

GFP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

264-449-2, monoclonal

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μg

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

The Green Fluorescent Protein (GFP) from the jellyfish Aequorea victoria is used as a fluorescent indicator for monitoring gene expression in a variety of cellular systems, including living organisms and fixed tissues. Unlike other bioluminescent reporters, GFP fluoresces in the absence of substrates, cofactors, or other intrinsic or extrinsic proteins. Purified GFP is a 27 kDa monomer consisting of 238 amino acids and emits green light (emission maximum at 509 nm) when excited with blue or UV light.

Specificity

Directed against a bacterially-expressed fusion protein. Also reacts with red-shifted GFP-variant. Does not react with enhanced GFP.

Immunogen

Epitope: original GFP only
highly-purified native GFP

Application

Anti-Green Fluorescent Protein Antibody, original GFP only, clone 264-449-2 is an antibody against Green Fluorescent Protein for use in IP, WB & IC.
Immunocytochemistry

Immunoblotting: 1:1000

Immunoprecipitation

Optimal working dilutions and protocols must be determined by the end user.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags

Quality

Tested

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02M Phosphate buffer, 0.25M NaCl, pH 7.6 with 0.1% sodium azide.

Storage and Stability

Store at 2-8°C for up to 12 months.

During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fei Mao et al.
Acta neuropathologica, 142(4), 629-642 (2021-06-22)
The neurodegenerative diseases amyotrophic lateral sclerosis (ALS) and frontotemporal lobar degeneration with TAR DNA-binding protein-43 (TDP-43) inclusions (FTLD-TDP) share the neuropathological hallmark of aggregates of TDP-43. However, factors governing the severity and regional distribution of TDP-43 pathology, which may account
Guoqiang Cai et al.
The FEBS journal, 272(7), 1625-1638 (2005-03-30)
Neurotransmitter transporters play a major role in achieving low concentrations of their respective transmitter in the synaptic cleft. The GABA transporter GAT1 belongs to the family of Na(+)- and Cl(-)-coupled transport proteins which possess 12 putative transmembrane domains and three
Alpha-synuclein-induced aggregation of cytoplasmic vesicles in Saccharomyces cerevisiae.
Soper, JH; Roy, S; Stieber, A; Lee, E; Wilson, RB; Trojanowski, JQ; Burd, CG; Lee, VM
Molecular Biology of the Cell null
Nguyet Nguyen et al.
Cell communication and signaling : CCS, 15(1), 48-48 (2017-11-16)
Human hyaluronic acid (HA) molecules are synthesized by three membrane spanning Hyaluronic Acid Synthases (HAS1, HAS2 and HAS3). Of the three, HAS1 is found to be localized more into the cytoplasmic space where it synthesizes intracellular HA. HA is a
Daniel Rodríguez-Martínez et al.
PloS one, 12(1), e0170776-e0170776 (2017-01-26)
Cryopreservation protocols are essential for stem cells storage in order to apply them in the clinic. Here we describe a new standardized cryopreservation protocol for GABAergic neural precursors derived from the medial glanglionic eminence (MGE), a promising source of GABAergic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service