Skip to Content
Merck
All Photos(1)

Key Documents

MAB2057

Sigma-Aldrich

Anti-Integrin α3 Antibody, clone ASC-6

clone ASC-6, Chemicon®, from mouse

Synonym(s):

CD49c

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€446.00

€446.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€446.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€446.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

ASC-6, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA3(3675)

Specificity

Human alpha3 integrin (CD49c)

Immunogen

Human squamous cell carcinoma cells (SCC9)

Application

Anti-Integrin α3 Antibody, clone ASC-6 is an antibody against Integrin α3 for use in FC, IP, IH.
Inhibition of cell attachment: inhibits SCC9 cell adhesion to human and EHS laminin, but not laminin peptide GD-2. Potentiates inhibition of SKOV-3 cell adhesion to EHS laminin by anti-integrin alpha 5 antibody GoH3. Does not inhibit either cell′s adhesion to fibronectin or collagen type IV.

Immunohistochemistry: on acetone-fixed frozen human epithelial tissues including tongue,foreskin and ovary. Not for use on formalin-fixed tissue.

Flow cytometry: stains human squamous cell carcinoma (SCC9), ovarian carcinoma (SKOV-3) and umbilical vein endothelial cells (HUVEC).

Working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Physical form

Format: Purified
Liquid in 0.02M PB pH 7.6, 0.25M NaCl containing 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi I Wu et al.
The Biochemical journal, 407(2), 171-177 (2007-07-26)
The transmembrane collagenase MT1-MMP (membrane-type 1 matrix metalloproteinase), also known as MMP-14, has a critical function both in normal development and in cancer progression, and is subject to extensive controls at the post-translational level which affect proteinase activity. As zymogen
Yu-Chen Gu et al.
Blood, 101(3), 877-885 (2002-10-24)
Laminins are alphabetagamma heterotrimeric extracellular proteins that regulate cellular functions by adhesion to integrin and nonintegrin receptors. Laminins containing alpha4 and alpha5 chains are expressed in bone marrow, but their interactions with hematopoietic progenitors are unknown. We studied human bone
Andrey Makarov et al.
Cell biology international, 37(10), 1080-1088 (2013-05-21)
Glioblastoma is the most common brain cancer. Ephrins and their Eph receptors play important roles in the development of central nervous system and the regulation of cancer cell migration and invasion. In a search for the Eph receptor complexes, we
Michelle B Chen et al.
Cancer research, 76(9), 2513-2524 (2016-03-19)
Tumor integrin β1 (ITGB1) contributes to primary tumor growth and metastasis, but its specific roles in extravasation have not yet been clearly elucidated. In this study, we engineered a three-dimensional microfluidic model of the human microvasculature to recapitulate the environment
Beta1 integrin mediates internalization of mammalian reovirus.
Maginnis, MS; Forrest, JC; Kopecky-Bromberg, SA; Dickeson, SK; Santoro, SA; Zutter et al.
Journal of virology null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service