Skip to Content
Merck
All Photos(1)

Documents

MAB2056Z

Sigma-Aldrich

Anti-Integrin α3 Antibody, clone ASC-1 (Azide Free)

clone ASC-1, Chemicon®, from mouse

Synonym(s):

CD49c

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

ASC-1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA3(3675)

Specificity

Human alpha3 integrin (CD49c)

Immunogen

Human squamous cell carcinoma cells (SCC9)

Application

Inhibition (Functional Activity): A previous lot of this antibody was shown to inhibit cell attachment by inhibiting SCC9 cell adhesion to human and EHS laminin, and laminin GD-2 peptide and potentiates the inhibition of SKOV-3 cell adhesion to EHS laminin by anti-integrin alpha 5 antibody GoH3. This antibody does not inhibit cell adhesion to fibronectin or collagen type IV.

Immunohistochemistry Analysis: A previous lot of this antibody was used on acetone-fixed frozen human epithelial tissues including tongue, foreskin and ovary. Not for use on formalin-fixed tissue.

Flow Cytometry Analysis: A previous lot of this antibody stains human squamous cell carcinoma (SCC9), ovarian carcinoma (SKOV-3) and umbilical vein endothelial cells (HUVEC).

Immunoprecipitation Analysis: A previous lot of this antibody precipitates PAGE bands of 130 kDa (reduced) or 116 kDa and 135 kDa (non-reduced) from surface-biotinylated SCC9 or SKOV-3 cells.
Anti-Integrin α3 Antibody, clone ASC-1 (Azide Free) inhibits cell attachment & detects level of Integrin α3 & has been published & validated for use in FC, IP, IH, Inhibition Assays.
Research Category
Cell Structure
Research Sub Category
Integrins

Target description

116 kDa

Physical form

Format: Purified
Purified antibody in PBS without preservatives.

Storage and Stability

Store at -20°C in undiluted aliquots.
Avoid repeated freeze-thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Kim et al.
The Journal of biological chemistry, 275(40), 30907-30915 (2000-07-25)
betaig-h3 is a transforming growth factor-beta-inducible cell adhesion molecule that has four characteristic homologous repeated domains. We made recombinant betaig-h3 proteins, which were highly active in mediating human corneal epithelial (HCE) cell adhesion and spreading. The 2nd and the 4th
Daniela I Staquicini et al.
Med (New York, N.Y.), 2(3), 321-342 (2021-04-20)
The extensive alveolar capillary network of the lungs is an attractive route for administration of several agents. One key functional attribute is the rapid onset of systemic action due to the absence of first-pass metabolism. Here we applied a combinatorial
Nick J I Hamilton et al.
The European respiratory journal, 55(6) (2020-05-24)
Current methods to replace damaged upper airway epithelium with exogenous cells are limited. Existing strategies use grafts that lack mucociliary function, leading to infection and the retention of secretions and keratin debris. Strategies that regenerate airway epithelium with mucociliary function

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service