Skip to Content
Merck
All Photos(1)

Documents

MAB201B

Sigma-Aldrich

Mouse Anti-Rabbit light chain Antibody, biotin conjugate

Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody product type

secondary antibodies

clone

monoclonal

species reactivity

rabbit

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

shipped in

wet ice

target post-translational modification

unmodified

General description

Antibody molecules typically comprise two immunoglobulin light chains covalently bound to a pair of heavy chains. Immunoglobulin light chains occur in two types, designated by the Greek letters kappa and lambda. Kappa and gamma light chains are approximately 250 amino acids in length with an average mass of about 25 kDa. The ratio of kappa to lambda found in the immunoglobulin population varies by species.

Specificity

Minimal cross-reaction with bovine, goat, Armenian hamster, horse, human, mouse, rat, and sheep.

Application

Mouse anti-Rabbit light chain Antibody, biotin conjugate is an antibody against Rabbit light chain for use in ELISA, FC, WB, IH.
Western Blotting: 1:20,000 - 1:400,000
Immunohistochemistry: 1:500 - 1:5,000 on tissue sections
Flow Cytometry: 1:200 - 1:1,000

Optimal working dilutions must be determined by the end user.

Target description

25 kDa

Physical form

Lyophilized from 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6, 0.05% Sodium Azide. 15 mg/mL BSA as stabilizer (IgG-Free, Protease-Free)

Storage and Stability

Maintain lyophilized product at 2°-8°C for up to 12 months. After reconstitution the product is stable for six weeks at 2°-8°C in the dark as an undiluted liquid. Reconstitute vial with of distilled water (addition of 0.5 ml water will yield a final concentration of 1.1 mg/mL). For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of protein (and buffer salts) will decrease to one-half of the original after the addition of glycerol. Avoid repeated freeze thaw cycles. Keep away from light.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Rampersaud et al.
British journal of pharmacology, 167(7), 1467-1479 (2012-07-11)
BACKGROUND AND PURPOSE Parkinson's disease (PD) is characterized by progressive dopaminergic cell loss; however, the noradrenergic system exhibits degeneration as well. Noradrenergic deficit in PD may be responsible for certain non-motor symptoms of the pathology, including psychiatric disorders and cognitive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service