Skip to Content
Merck
All Photos(1)

Key Documents

MAB1973Z

Sigma-Aldrich

Anti-Integrin α1 Antibody, clone FB12, azide free

clone FB12, Chemicon®, from mouse

Synonym(s):

CD49a, MAB1973

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€586.00

€586.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€586.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€586.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

FB12, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA1(3672)

Specificity

Reacts with the I domain (Val151 - Ala364) of human alpha1 integrin (Fabbri, 1996). Precipitates proteins of Mr 180 kDa, pI 5.9-6.0 (alpha chain) and Mr 115 kDa, pI 4.5-5.0 (beta chain) from human lymphocyte lysates (Zocchi, 1991). Does not recognize recombinant I domain of rat integrin alpha1.

Application

Detect Integrin α1 using this Anti-Integrin α1 Antibody, clone FB12, azide free validated for use in ELISA, FC, IH, IP, FUNC & WB.
Immunohistochemistry: 2 μg/mL, with paraformaldehyde fixation. Immunoblotting: 2 μg/mL

Immunoprecipitation: 1 μg/mL

FACS analysis: 2 μg/mL

ELISA: 0.5 μg/mL

Inhibition of activated human lymphocyte binding to laminin, collagen IV and fibronectin (Fabbri, 1996): 1-5 μg/mL

Optimal working dilutions must be determined by end user.

Target description

130 kDa

Physical form

Format: Purified
Purified immunoglobulin in 0.02M PB, pH 7.6, 0.25M NaCl.

Analysis Note

Control
Microvascular endothelium

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuhiro Katayama et al.
International journal of molecular sciences, 21(6) (2020-03-19)
Keloids are dermal fibroproliferative tumors that arise beyond the boundary of the original wound edges and invades adjacent tissue. Keloids are characterized by the extensive production of extracellular matrix (ECM) and abnormal fibroblast proliferation. Chondroitin sulfate (CS) is one of
Tobias Gerber et al.
Oncotarget, 8(1), 846-862 (2016-12-03)
Recent technological advances in single-cell genomics make it possible to analyze cellular heterogeneity of tumor samples. Here, we applied single-cell RNA-seq to measure the transcriptomes of 307 single cells cultured from three biopsies of three different patients with a BRAF/NRAS
Shawn M Sweeney et al.
The Journal of biological chemistry, 278(33), 30516-30524 (2003-06-06)
Angiogenesis depends on proper collagen biosynthesis and cross-linking, and type I collagen is an ideal angiogenic scaffold, although its mechanism is unknown. We examined angiogenesis using an assay wherein confluent monolayers of human umbilical vein endothelial cells were overlain with
Integrin upregulation and localization to focal adhesion sites in pregnant human myometrium.
Burkin, HR; Rice, M; Sarathy, A; Thompson, S; Singer, CA; Buxton, IL
Reproductive Sciences null
Christopher L Gilchrist et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 25(6), 829-840 (2007-02-24)
Cellular interactions with the extracellular matrix are key factors regulating cell survival, differentiation, and response to environmental stimuli in cartilagenous tissues. Much is known about the extracellular matrix proteins in the intervertebral disc (IVD) and their variations with region, age

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service