Skip to Content
Merck
All Photos(1)

Key Documents

MAB1921P

Sigma-Aldrich

Anti-Laminin beta 1 (LAMB1) Antibody

CHEMICON®, mouse monoclonal, 4E10

Synonym(s):

Laminin B1 Chain

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€459.00

€459.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€459.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€459.00


Check Cart for Availability

Request a Bulk Order

Product Name

Anti-Laminin β1 Antibody, clone 4E10, clone 4E10, Chemicon®, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4E10, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LAMB1(3912)

Specificity

Reacts with the beta1 chain of intact laminin and blocks the neurite stimulating activity of human laminin. No cross-reactivity with mouse laminin.

Immunogen

Purified human laminin

Application

Detect Laminin β1 using this Anti-Laminin β1 Antibody, clone 4E10 validated for use in ELISA, IF, IP & WB.
Research Category
Cell Structure
Research Sub Category
ECM Proteins
Western blot

Immunoprecipitation

Immunoflurescence

ELISA

Immunoaffinity chromatography

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in PBS, pH 7.6, containing 0.1% sodium azide.

Storage and Stability

Maintain at 2°-8°C for up to 12 months from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Fan et al.
Frontiers in oncology, 11, 740410-740410 (2021-12-21)
Craniopharyngiomas (CPs) predominantly involving the third ventricle were commonly termed "intraventricular" lesions. The aim of this study was to clarify the anatomical relationship between the tumor and the third ventricle by both surgical and histological investigation. A retrospective review of
Radhika Mhatre et al.
Annals of Indian Academy of Neurology, 24(2), 198-203 (2021-07-06)
Immunocharacterization of congenital muscular dystrophy (CMD) to determine the frequency of various subtypes in a large Indian Cohort. This retrospective (2014-2017) study was carried on muscle biopsies of clinically suspected cases of CMD with histological evidence of dystrophy/myopathic features. Immunohistochemistry
Dongsheng Guo et al.
eLife, 11 (2022-01-26)
Skeletal muscle myoblasts (iMyoblasts) were generated from human induced pluripotent stem cells (iPSCs) using an efficient and reliable transgene-free induction and stem cell selection protocol. Immunofluorescence, flow cytometry, qPCR, digital RNA expression profiling, and scRNA-Seq studies identify iMyoblasts as a
Amber N Stratman et al.
Blood, 114(24), 5091-5101 (2009-10-14)
We show that endothelial cell (EC)-generated vascular guidance tunnels (ie, matrix spaces created during tube formation) serve as conduits for the recruitment and motility of pericytes along EC ablumenal surfaces to facilitate vessel maturation events, including vascular basement membrane matrix

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service