Skip to Content
Merck
All Photos(1)

Documents

MAB1763

Sigma-Aldrich

Anti-Fibulin-3 Antibody

ascites fluid, clone G3-5, Chemicon®

Synonym(s):

EFEMP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

G3-5, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

Gene Information

human ... EFEMP1(2202)

General description

Fibulin-3 is a protein that belongs to a family of extracellular proteins expressed in the basement membranes of blood vessels and the gene that encodes fibulin 3 is EFEMP1. It is a secreted protein that is present in serum, urine and tissue samples. A single mutation in FBLN3 is responsible for an autosomal dominant form of macular degeneration (Marmorstein, et al., 2002)

Specificity

Clone G3-5 recognizes human fibulin-3 with a MW of 55 kDa.

Immunogen

Epitope: amino acids 107-493
GST-tagged fibulin-3 fusion protein containing amino acids 107-493.

Application

Western blot: 1:100 after immunoprecipitation
Research Category
Cell Structure
Research Sub Category
ECM Proteins
This Anti-Fibulin-3 Antibody is validated for use in WB for the detection of Fibulin-3.

Physical form

Ascites containg no preservatives.

Storage and Stability

Maintain at -20°C for up to 12 months from date of receipt in undiluted aliquots. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Positive: ARPE-19 cell line.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service