Skip to Content
Merck
All Photos(1)

Key Documents

MAB1759F

Sigma-Aldrich

Anti-CD59 Antibody, clone 2/24, FITC conjugated

clone 2/24, Chemicon®, from mouse

Synonym(s):

Protectin

Sign Into View Organizational & Contract Pricing

Select a Size

100 ASSAYS
€395.00

€395.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 ASSAYS
€395.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

€395.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2/24, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunofluorescence: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD59(966)

General description

BIOCHEMISTRY:

p I = 6.1

Specificity

This antibody binds to a 17-25kD glycoprotein (Fletcher et al., 1990a,b) which is distributed on human cells and has a glycosyl-phosphatidylinositol (GPI) linkage to the plasma membrane(Hadam, 1989). It is identical to other CD59 monoclonals, MEM-43 and YTH 53.1, defined at the 4th International Conference on Human Leucocyte Differentiation Antigens(Davies et al., 1989; Stefanova et al., 1989). CD59 is an inhibitor of attack by autologous complement(Ojcius et al., 1990) possibly by limiting incorporation of C9 into the membrane complex(Rollins & Sims, 1990; Meri et al., 1990). GPI linked proteins such as CD59 are deficient on cells from patients with the acquired clonal disorder paroxysmal nocturnal haemoglobulinuria (PNH) in which the erythrocytes are susceptible to lysis by complement and leading to anemia(Rosse, 1990). Evidence suggests that CD59 may be the most important complement regulatory protein missing in PNH(Fletcher et al., 1990; Yamashina et al., 1990).

Cell reactivity (Fletcher et al., 1990a,b; Hadam, 1989):

The antibody reacts with all leucocytes, platelets, fibroblasts, erythrocytes and most cell lines. It is negative on U937 and Daudi cell lines. It is reduced on affected cells from patients with PNH with a distribution reflecting the clonal nature of the disease.

Immunogen

Human erythrocytes

Application

Anti-CD59 Antibody, clone 2/24, FITC conjugated detects level of CD59 & has been published & validated for use in FC & IF.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This monoclonal is useful in studies of complement and homologous restriction of complement action of the properties of GPI linked proteins and in the characterisation of cells from patients with PNH.

SUGGESTED USAGE:
Flow cytometry and immunofluorescence – use 10µL direct from the vial per 1x106 cells in a volume of 100 µL.
For analysis of erythrocytes, whole blood samples should be diluted approximately 500 fold.
For nucleated cells, use of the buffer coat will remove the burden of erythrocytes.
Platelets may be harvested by low speed centrifugation.

Physical form

Purified immunoglobulin. FITC Conjugated. The antibody is supplied in phosphate buffered saline, pH 7.4, containing 0.2% bovine serum albumin and 0.1% sodium azide. The characteristics of each lot are tested by electrophoresis and flow cytometry.

Storage and Stability

STORAGE

Store at 2 to 8oC, for up to 6 months. For prolonged periods, store below -20oC in undiluted aliquots. AVOID REPEATED FREEZE/THAW CYCLES.

WARNING

The monoclonal reagent solution contains 0.1% sodium azide as a preservative. Due to potential hazards arising from the build up of this material in pipes, spent reagent should be disposed of with liberal volumes of water.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service