Skip to Content
Merck
All Photos(1)

Key Documents

MAB1553

Sigma-Aldrich

Anti-Titin Antibody, clone 9B9

culture supernatant, clone 9B9, Chemicon®

Synonym(s):

Anti-CMD1G, Anti-CMPD4, Anti-EOMFC, Anti-HMERF, Anti-LGMD2J, Anti-LGMDR10, Anti-MYLK5, Anti-SALMY, Anti-TMD

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€528.00

€528.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
1 ML
€528.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€528.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

9B9, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TTN(7273)

Specificity

Human skeletal muscle titin.

Immunogen

Electrophoretically purified human skeletal muscle titin.

Application

Immunochemistry: undiluted to 1:10

Western blotting: 1:10-1:20

Optimal dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton
This Anti-Titin Antibody, clone 9B9 is validated for use in WB, IH for the detection of Titin.

Physical form

Liquid. Contains 0.1% sodium azide.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rafael De Cid et al.
Neurology, 85(24), 2126-2135 (2015-11-20)
To identify the genetic defects present in 3 families with muscular dystrophy, contractures, and calpain 3 deficiency. We performed targeted exome sequencing on one patient presenting a deficiency in calpain 3 on Western blot but for which mutations in the
Hong-Kee Tan et al.
Stem cells translational medicine, 3(5), 586-598 (2014-03-22)
Induced pluripotent stem cells (iPSCs) derived from somatic cells of patients can be a good model for studying human diseases and for future therapeutic regenerative medicine. Current initiatives to establish human iPSC (hiPSC) banking face challenges in recruiting large numbers
Stoyan Petkov et al.
Cells, 9(11) (2020-11-11)
Induced pluripotent stem cells (iPSCs) hold enormous potential for the development of cell-based therapies; however, the safety and efficacy of potential iPSC-based treatments need to be verified in relevant animal disease models before their application in the clinic. Here, we
Michael Stauske et al.
Cells, 9(6) (2020-06-04)
Non-human primates (NHP) are important surrogate models for late preclinical development of advanced therapy medicinal products (ATMPs), including induced pluripotent stem cell (iPSC)-based therapies, which are also under development for heart failure repair. For effective heart repair by remuscularization, large

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service