Skip to Content
Merck
All Photos(2)

Key Documents

MAB1233-I

Sigma-Aldrich

Anti-Estradiol (6-BSA) Antibody

purified antibody, clone 6E1, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€153.00
100 μL
€383.00

€153.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€153.00
100 μL
€383.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€153.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

6E1, monoclonal

species reactivity

human

packaging

antibody small pack of 100 μL

technique(s)

ELISA: suitable
activity assay: suitable
immunocytochemistry: suitable

isotype

IgG

shipped in

dry ice

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Specificity

Clone 9E1 is a mouse monoclonal antibody that detects BSA-conjugated Estradiol.

Immunogen

BSA-conjugate Estradiol (Estradiol-6-BSA).

Application

Quality Control TestingEvaluated by Immunocytochemistry in OVCAR 3 cells.Immunocytochemistry Analysis: A 1:25 dilution of this antibody detected Estradiol (6-BSA) in OVCAR 3 cells.Tested ApplicationsELISA Analysis: Various titrations (2-fold serial dilutions) starting at 100 ng/mL from a representative lot detected Estradiol (6-BSA).Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Estradiol (6-BSA), clone 6E1, Cat. No. MAB1233-I, is a mouse monoclonal antibody that detects Estradiol-6-BSA and is tested for use in ELISA and Immunocytochemistry.

Target description

Estradiol is a major female sex hormone that regulates estrous and menstrual cycle. It is also present in smaller quantities in males where it is an active metabolic product of testosterone. In females, it is mainly produced by the granulosa cells in the follicles of ovaries. Smaller amounts of estradiol are also produced by the adrenal glands. It acts as an agonist for the estrogen receptor (ER) and can bind to both ERa and ERb. It is synthesized from cholesterol where androstenedione is a key intermediate which is converted to estradiol by aromatase. It is metabolized via hydroxylation into catechol estrogens. In the liver it can undergo hydroxylation into 2-hydoxyestradiol by CYP1A2, CYP3A4, and CYP2C9. Estradiol is reported to directly act on the central nervous system to modulate energy balance and reduced levels of estradiol following ovarian insufficiency have been linked to menopause, hyperphagia, and weight gain. (Ref.: Gonzales-Garcia, I., et al. (2018). Cell Rep. 25(2): 413-423).

Physical form

Purified mouse monoclonal antibody IgG in PBS without azide.

Storage and Stability

Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service