Skip to Content
Merck
All Photos(1)

Key Documents

MAB1219

Sigma-Aldrich

Anti-CD14 Antibody, clone 2D-15C

clone 2D-15C, Chemicon®, from mouse

Synonym(s):

LPS Receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€327.00

€327.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€327.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€327.00


Estimated to ship on31 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2D-15C, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD14(929)

Related Categories

General description

BIOCHEMISTRY: p I = 5.8

Specificity

This antibody belongs to CD14 (assigned by the Third International Workshop on Leucocyte Differentiation Antigens, Oxford, 1986; McMichael et al. Eds., 1986). and reacts with 50 to 55kD protein. It will detect blood monocytes, Kupffer cells, red pulp macrophages, dendritic cells and also "epithelioid" or giant cells within granulomatous tissue (Hancock et al., 1983), foam cells in renal interstitium (Nolasco et al., 1984) and placental dendritic cells and macrophages (Barnett et al., 1984; Brooks et al., 1983; Polli et al., 1984; Hopper et al., 1986). Monocytes react with this antibody in the early stages of differentiation, promonocytes and monoblasts do not. Activity is not lost on monocyte/macrophage activation (Hancock et al., 1983).



Cell reactivity

Reacts with peripheral blood monocytes and weakly with granulocytes. Negative to platelets, erythrocytes and lymphocytes. Sometimes positive for chronic myeloid leukemia (CML) and acute myeloid leukemia (AML). Negative to B-cell chronic lymphocytic leukemia (B-CLL), T-cell chronic lymphocytic leukemia (T-CLL) and common acute lymphoblastic leukemia (CALL).

Immunogen

Human peripheral blood mononuclear cells.

Application

Anti-CD14 Antibody, clone 2D-15C detects level of CD14 & has been published & validated for use in FC, IH.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This antibody is an important general cell marker for mononuclear phagocytes in normal samples and a variety of disease states including a proportion of cases of myeloid leukemia. It is suitable for flow cytometry, and immunoperoxidase staining on frozen tissue sections.

SUGGESTED USAGE DILUTION

1. Flow cytometry and indirect immunofluorescence 1:25



Dilute with isotonic buffer. Use 50 μl per 1 x 106 peripheral blood mononuclear cells (PBMC) in 100 μl buffer.



2. Indirect immunoperoxidase staining - the final dilution will depend on the assay conditions and detection system employed. However, a dilution of at least 1:25 will be applicable to most commonly used systems.

Physical form

Format: Purified
The purified antibody is supplied in phosphate buffered saline, pH 7.4, containing 0.2% bovine serum albumin and 0.1% sodium azide. The characteristics of each lot are tested by electrophoresis and flow cytometry.

Storage and Stability

Store at 2 to 8°C, for up to 6 months. For prolonged periods, store below -20°C in undiluted aliquots. AVOID REPEATED FREEZE/THAW CYCLES.

WARNING: The monoclonal reagent solution contains 0.1% sodium azide as a preservative. Due to potential hazards arising from the build up of this material in pipes, spent reagent should be disposed of with liberal volumes of water.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susan S Ishmael et al.
Journal of leukocyte biology, 87(2), 291-300 (2009-11-06)
In human basophils from different subjects, maximum IgE-mediated histamine release and the level of syk protein expression correlate well. It is not clear when in the basophil's lifetime the set-point for syk expression is reached or how expression levels are
Larissa M Macedo et al.
Journal of cellular physiology, 236(1), 366-378 (2020-06-11)
The effects of the renin-angiotensin system (RAS) on stem cells isolated from human dental apical papilla (SCAPs) are completely unknown. Therefore, the aim of this study was to identify RAS components expressed in SCAPs and the effects of angiotensin (Ang)
Andreas Weigert et al.
Nature communications, 13(1), 6078-6078 (2022-10-15)
Fibrocytes are bone marrow-derived monocytic cells implicated in wound healing. Here, we identify their role in lung cancer progression/ metastasis. Selective manipulation of fibrocytes in mouse lung tumor models documents the central role of fibrocytes in boosting niche features and
Arantxa Blázquez-Prunera et al.
Stem cell research & therapy, 8(1), 103-103 (2017-04-30)
Mesenchymal stem cells (MSCs) show promising characteristics for their use in advanced therapy medicinal products. However, there are some unresolved concerns, such as the use of animal components for their expansion. In this study we assessed the suitability of a
Giulia Detela et al.
Biotechnology journal, 13(2) (2018-01-16)
Human mesenchymal stromal cells (hMSCs) are excellent candidates for cell therapy but their expansion to desired clinical quantities can be compromised by ex vivo processing, due to differences between donor material and process variation. The aim of this article is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service